Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-17 |
---|---|
miRBase Accession Number: | MI0000071 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAAAGUGCUUACAGUGCAGGUAG |
Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Callithrix jacchus, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pig, Pteropus alecto, Rabbit, Salmo salar, Taeniopygia guttata, Tupaia chinensis |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUUACAGUGCAGGUAG |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-17-5p | MIMAT0000070 | |
Mouse | mmu-miR-17-5p | MIMAT0000649 | |
Rat | rno-miR-17-5p | MIMAT0000786 | |
Alligator mississippiensis | ami-miR-17-5p | MIMAT0038047 | |
Anolis carolinensis | aca-miR-17-5p | MIMAT0021803 | |
Callithrix jacchus | cja-miR-17 | MIMAT0039391 | |
View More Rows | |||
Cricetulus griseus | cgr-miR-17-5p | MIMAT0023795 | |
Cyprinus carpio | ccr-miR-17-5p | MIMAT0026236 | |
Dasypus novemcinctus | dno-miR-17-5p | MIMAT0047509 | |
Guinea pig | cpo-miR-17-5p | MIMAT0046857 | |
Horse | eca-miR-106a | MIMAT0013194 | |
Horse | eca-miR-17 | MIMAT0013084 | |
Microcebus murinus | mmr-miR-17 | MIMAT0049198 | |
Nomascus leucogenys | nle-miR-17 | MIMAT0049455 | |
Ornithorhynchus anatinus | oan-miR-17-5p | MIMAT0007278 | |
Otolemur garnettii | oga-miR-17 | MIMAT0049648 | |
Pig | ssc-miR-17-5p | MIMAT0007755 | |
Pteropus alecto | pal-miR-17-5p | MIMAT0039959 | |
Rabbit | ocu-miR-17-5p | MIMAT0048109 | |
Salmo salar | ssa-miR-17-5p | MIMAT0032414 | |
Taeniopygia guttata | tgu-miR-17a-5p | MIMAT0017390 | |
Tupaia chinensis | tch-miR-17-5p | MIMAT0036516 |