Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-20b-5p |
---|---|
miRBase Accession Number: | MI0001519 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAAAGUGCUCAUAGUGCAGGUAG |
Species: | Human, Mouse, Rat, Callithrix jacchus, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rhesus monkey, Taeniopygia guttata, Xenopus laevis, Xenopus tropicalis |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUCAUAGUGCAGGUAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-20b-5p | MIMAT0001413 |
Mouse | mmu-miR-20b-5p | MIMAT0003187 |
Rat | rno-miR-20b-5p | MIMAT0003211 |
Callithrix jacchus | cja-miR-20b | MIMAT0039389 |
Chicken | gga-miR-20b-5p | MIMAT0001411 |
Columba livia | cli-miR-20b-5p | MIMAT0038434 |
View More Rows | ||
Cricetulus griseus | cgr-miR-20b | MIMAT0041172 |
Dasypus novemcinctus | dno-miR-20b-5p | MIMAT0047522 |
Gorilla gorilla | ggo-miR-20b | MIMAT0024227 |
Horse | eca-miR-20b | MIMAT0013202 |
Monodelphis domestica | mdo-miR-20b-5p | MIMAT0028764 |
Ornithorhynchus anatinus | oan-miR-20b-5p | MIMAT0006853 |
Pan troglodytes | ptr-miR-20b | MIMAT0008069 |
Pongo pygmaeus | ppy-miR-20b | MIMAT0015732 |
Pteropus alecto | pal-miR-20b-5p | MIMAT0040005 |
Python bivittatus | pbv-miR-20b-5p | MIMAT0038847 |
Rhesus monkey | mml-miR-20b-5p | MIMAT0006164 |
Taeniopygia guttata | tgu-miR-20b-5p | MIMAT0014591 |
Xenopus laevis | xla-miR-20 | MIMAT0001348 |
Xenopus tropicalis | xtr-miR-20b | MIMAT0003707 |