Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127394.3 | NP_001120866.1 | 4 | 1669 | ||
NM_001300764.1 | NP_001287693.1 | 5 | 1823 | |||
NM_001300766.1 | NP_001287695.1 | 5 | 1707 | |||
NM_052965.3 | NP_443197.1 | 5 | 1811 | |||
NR_023349.2 | 6 | 1928 | ||||
NR_125335.1 | 6 | 1891 | ||||
GenBank mRNA | AF288394.1 | - | 5 | 1786 | ||
AK098196.1 | - | 5 | 1708 | |||
BC022030.1 | - | 5 | 1756 | 4824025 | ||
BG285158.1 | - | 1 | 407 | |||
BM789892.1 | - | 1 | 532 |