Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320108.1 | NP_001307037.1 | 2 | 2309 | ||
NM_003708.4 | NP_003699.3 | 3 | 2568 | |||
GenBank mRNA | AB209332.1 | - | 2 | 2309 | ||
AF057034.1 | - | 3 | 792 | |||
AF086735.1 | - | 3 | 1508 |