Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005401.4 | NP_005392.2 | 17, 18 | 3923 | ||
XM_017001941.1 | XP_016857430.1 | 18, 19 | 4010 | |||
GenBank mRNA | AK291641.1 | - | 17, 18 | 3855 | ||
BC101754.1 | - | 16, 17 | 3330 | 8069260 | ||
BC104803.1 | - | 16, 17 | 3330 | 8143806 | ||
HQ116786.1 | - | 15, 16 | 3182 | |||
X82676.1 | - | 17, 18 | 3464 |