Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 26 | 2710 | ||
XM_011545742.2 | XP_011544044.1 | 29 | 2920 | |||
XM_011545744.2 | XP_011544046.1 | 14 | 1520 | |||
XM_011545745.1 | XP_011544047.1 | 12 | 1288 | |||
XM_011545746.2 | XP_011544048.1 | 12 | 1391 | |||
XM_017022947.1 | XP_016878436.1 | 30 | 2993 | |||
XM_017022948.1 | XP_016878437.1 | 13 | 2092 | |||
XM_017022949.1 | XP_016878438.1 | 11 | 2363 | |||
GenBank mRNA | AK128606.1 | - | 15 | 1578 | ||
BC038400.1 | - | 3 | 155 | |||
BC109294.1 | - | 15 | 1485 | |||
BC109295.1 | - | 14 | 1438 |