Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005381.2 | NP_001005381.2 | 7 | 586 | ||
XM_008772490.1 | XP_008770712.1 | 372 | ||||
GenBank mRNA | AY303540.1 | - | 604 | |||
BC079094.1 | - | 7 | 557 | 7123989 | ||
CB582878.1 | - | 7 | 586 |