Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tGap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007635.2 | NP_001007636.2 | 16 | 2195 | ||
XM_006232134.2 | XP_006232196.1 | 18 | 2537 | |||
GenBank mRNA | AY631396.1 | - | 16 | 2195 | ||
AY631398.1 | - | 9 | 1339 | |||
AY631399.1 | - | 16 | 1960 | |||
AY631400.1 | - | 17 | 2162 |