Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005879.2 | NP_005870.2 | 2, 3 | 270 | ||
XM_011533264.1 | XP_011531566.1 | 2, 3 | 275 | |||
XM_017005526.1 | XP_016861015.1 | 2, 3 | 295 | |||
GenBank mRNA | AK225873.1 | - | 2, 3 | 246 | ||
AK292172.1 | - | 2, 3 | 258 | |||
AK295780.1 | - | 2, 3 | 275 | |||
BC000310.2 | - | 2, 3 | 251 | 2821007 | ||
BC019283.1 | - | 2, 3 | 251 | 2821007 | ||
BX473682.1 | - | 2, 3 | 278 | |||
U77845.1 | - | 2, 3 | 266 |