Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNEP1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281789.1 | NP_001268718.1 | 6 | 1505 | ||
NM_153261.5 | NP_694993.2 | 7 | 1556 | |||
NR_104042.1 | 6 | 1482 | ||||
NR_104043.1 | 5 | 1431 | ||||
GenBank mRNA | AF086322.1 | - | 1 | 67 | ||
AK095420.1 | - | 6 | 1478 | |||
AK307132.1 | - | 6 | 1553 | |||
BC022550.1 | - | 5 | 1381 | |||
BC036683.2 | - | 7 | 1485 | 5266084 | ||
CR933600.1 | - | 7 | 1502 | |||
FJ516381.1 | - | 6 | 1433 |