Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003463.4 | NP_003454.1 | 5 | 1525 | ||
XM_011536111.1 | XP_011534413.1 | 5 | 864 | |||
XM_011536112.1 | XP_011534414.1 | 5 | 1623 | |||
XM_017011270.1 | XP_016866759.1 | 5 | 881 | |||
XM_017011271.1 | XP_016866760.1 | 5 | 861 | |||
XM_017011272.1 | XP_016866761.1 | 5 | 1525 | |||
GenBank mRNA | AB451314.1 | - | 4 | 372 | ||
AB451444.1 | - | 4 | 372 | |||
AK312526.1 | - | 5 | 1009 | |||
BC023975.2 | - | 5 | 967 | 2960001 | ||
BC045571.1 | - | 5 | 1525 | 4826233 | ||
CR749458.1 | - | 7 | 1359 | |||
U48296.1 | - | 4 | 764 | |||
U69701.1 | - | 2 | 136 |