Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163521.1 | NP_001156993.1 | 10 | 1486 | ||
NM_028504.1 | NP_082780.1 | 10 | 1519 | |||
GenBank mRNA | AK006591.1 | - | 10 | 1520 | ||
BC137683.1 | - | 10 | 1443 | 8860702 | ||
BC144828.1 | - | 10 | 1410 | 9053376 |