Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015774.3 | NP_056589.1 | 16 | 1726 | ||
GenBank mRNA | AK009667.1 | - | 6 | 951 | ||
AK142595.1 | - | 16 | 1717 |