Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172707.3 | NP_766295.2 | 8 | 2535 | ||
GenBank mRNA | AK004686.2 | - | 8 | 2246 | ||
AK088893.1 | - | 8 | 2175 | |||
AK147112.1 | - | 8 | 2338 | |||
AK160743.1 | - | 8 | 2244 | |||
AK166168.1 | - | 8 | 2454 | |||
AK166416.1 | - | 8 | 2541 | |||
AK166623.1 | - | 8 | 2438 | |||
AK168141.1 | - | 8 | 2431 | |||
AK168512.1 | - | 8 | 2182 | |||
AK169379.1 | - | 8 | 2284 | |||
AK171283.1 | - | 8 | 2404 | |||
BC046832.1 | - | 8 | 2235 | 6812937 |