Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242407.1 | NP_001229336.1 | 3 | 359 | ||
NM_175392.3 | NP_780601.1 | 3 | 527 | |||
GenBank mRNA | AK040713.1 | - | 2 | 1417 | ||
AK077618.1 | - | 3 | 320 | |||
AK140122.1 | - | 3 | 345 | |||
AK154200.1 | - | 3 | 348 | |||
AK165996.1 | - | 3 | 350 | |||
BC051404.1 | - | 3 | 527 | 6395030 |