Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001841.2 | NP_001832.1 | 2 | 745 | ||
XM_011540629.2 | XP_011538931.1 | 2 | 1028 | |||
XM_017000261.1 | XP_016855750.1 | 5 | 1720 | |||
GenBank mRNA | AJ430063.1 | - | 1 | 654 | ||
AM156854.1 | - | 1 | 618 | |||
AM156855.1 | - | 1 | 618 | |||
AM156856.1 | - | 1 | 618 | |||
BC069722.1 | - | 2 | 687 | 7262528 | ||
BC074767.2 | - | 1 | 646 | 30915186 | ||
BC095545.1 | - | 1 | 615 | |||
EU517121.1 | - | 3 | 877 | |||
X74328.1 | - | 2 | 744 |
Set Membership: |
![]() |