Gene Symbol: PCDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040429.2 | NP_001035519.1 | 4 | 4127 | ||
XM_005266357.2 | XP_005266414.1 | 5 | 5572 | |||
XM_005266358.2 | XP_005266415.1 | 4 | 5675 | |||
GenBank mRNA | AF029343.1 | - | 4 | 3980 | ||
AK289980.1 | - | 4 | 3970 | |||
AK307545.1 | - | 5 | 1861 | |||
BC028165.1 | - | 4 | 3261 | 5245587 |