Gene Symbol: Gspt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008179.2 | NP_032205.2 | 1 | 925 | ||
GenBank mRNA | AB003503.1 | - | 1 | 678 | ||
AK019241.1 | - | 1 | 919 | |||
BC117825.1 | - | 1 | 832 | 40092874 | ||
BC117826.1 | - | 1 | 832 | 40092877 |