Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dixdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178118.2 | NP_835219.1 | 18 | 2026 | ||
GenBank mRNA | AK082960.1 | - | 1 | 55 | ||
AK129432.1 | - | 14 | 1206 | |||
AK147565.1 | - | 12 | 2766 | |||
AK147596.1 | - | 12 | 2766 | |||
AY549883.1 | - | 18 | 1884 | |||
AY549884.1 | - | 18 | 1887 | |||
AY549885.1 | - | 14 | 1176 | |||
AY549886.1 | - | 14 | 1185 | |||
AY549887.1 | - | 12 | 927 | |||
BC048182.1 | - | 18 | 2017 | 6826299 | ||
BC063085.1 | - | 18 | 2017 | 6826299 |