Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081205.1 | NP_001074674.1 | 6 | 1008 | ||
GenBank mRNA | AK014427.1 | - | 6 | 961 | ||
AK035018.1 | - | 1 | 100 | |||
AK075654.1 | - | 6 | 1012 | |||
AK163026.1 | - | 1 | 295 |