Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031655.1 | NP_113843.1 | 1 | 264 | ||
GenBank mRNA | BC081763.1 | - | 1 | 250 | 7125782 | |
FQ220594.1 | - | 1 | 279 | |||
FQ229501.1 | - | 1 | 256 | |||
U40260.1 | - | 1 | 231 | |||
X76985.1 | - | 1 | 264 |