Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021748.1 | NP_068516.1 | 549 | |||
GenBank mRNA | AF089839.1 | - | 4 | 347 | ||
AF142097.1 | - | 549 | ||||
AF189019.1 | - | 549 | ||||
FQ211783.1 | - | 588 |