Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003965.2 | 323 | ||||
GenBank mRNA | AK131930.1 | - | 324 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ043634.1 | - | 3 | 324 | Chr. 9 - 62842027 - 62868000 [-] on Build GRCm38 |