Gene Symbol: Zfp637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177684.2 | NP_808352.1 | 3 | 245 | ||
GenBank mRNA | AB221671.1 | - | 2 | 148 | ||
AK146789.1 | - | 3 | 378 | |||
BC021601.1 | - | 3 | 274 | 5150384 | ||
BC037994.1 | - | 2 | 228 | |||
BC048843.1 | - | 3 | 245 | 5031082 |