Gene Symbol: Zbtb7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054002.1 | NP_446454.1 | 2 | 1156 | ||
XM_006240836.2 | XP_006240898.1 | 2 | 1803 | |||
XM_006240837.2 | XP_006240899.1 | 2 | 1493 | |||
XM_006240838.2 | XP_006240900.1 | 2 | 1381 | |||
GenBank mRNA | D88450.1 | - | 2 | 1156 | ||
FQ212204.1 | - | 2 | 1121 |