Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107095.1 | NP_001100565.1 | 9, 10 | 1622 | ||
GenBank mRNA | AB438101.1 | - | 12, 13 | 2146 |