Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003927.4 | NP_003918.1 | 2 | 917 | ||
NM_015832.4 | NP_056647.1 | 2 | 917 | |||
GenBank mRNA | AF039701.1 | - | 1 | 55 | ||
AF072242.1 | - | 2 | 917 | |||
AF072246.1 | - | 2 | 220 | |||
BC032638.1 | - | 2 | 758 | 5496721 |