Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 12 | 2730 | ||
NM_014903.5 | NP_055718.4 | 12 | 2730 | |||
XM_005269215.3 | XP_005269272.1 | 12 | 6731 | |||
XM_011538940.2 | XP_011537242.1 | 12 | 6732 | |||
XM_011538941.2 | XP_011537243.1 | 12 | 6731 | |||
XM_011538944.2 | XP_011537246.1 | 12 | 6729 | |||
XM_017020164.1 | XP_016875653.1 | 12 | 6733 | |||
XM_017020165.1 | XP_016875654.1 | 12 | 6733 | |||
XM_017020166.1 | XP_016875655.1 | 12 | 2719 | |||
XM_017020167.1 | XP_016875656.1 | 11 | 2650 | |||
XM_017020168.1 | XP_016875657.1 | 12 | 6732 | |||
XM_017020169.1 | XP_016875658.1 | 12 | 6731 | |||
XM_017020170.1 | XP_016875659.1 | 12 | 6728 | |||
XM_017020171.1 | XP_016875660.1 | 4 | 6026 | |||
XM_017020172.1 | XP_016875661.1 | 4 | 1664 | |||
XM_017020173.1 | XP_016875662.1 | 4 | 6024 | |||
XM_017020174.1 | XP_016875663.1 | 4 | 6024 | |||
XM_017020175.1 | XP_016875664.1 | 4 | 1903 | |||
GenBank mRNA | AF397731.1 | - | 11 | 2314 | ||
AJ488201.1 | - | 12 | 2602 |