Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175736.4 | NP_783863.4 | 16, 17 | 2042 | ||
NM_198900.2 | NP_944489.2 | 15, 16 | 1889 | |||
XM_005269218.2 | XP_005269275.1 | 16, 17 | 2021 | |||
XM_011538968.2 | XP_011537270.1 | 16, 17 | 2511 | |||
XM_011538969.2 | XP_011537271.1 | 16, 17 | 2510 | |||
XM_011538970.2 | XP_011537272.1 | 16, 17 | 2510 | |||
XM_011538971.2 | XP_011537273.1 | 16, 17 | 2086 | |||
XM_011538972.2 | XP_011537274.1 | 15, 16 | 2356 | |||
XM_011538973.2 | XP_011537275.2 | 12, 13 | 1501 | |||
XM_011538974.2 | XP_011537276.1 | 17, 18 | 1843 | |||
GenBank mRNA | AB095934.2 | - | 16, 17 | 2042 | ||
AK127200.1 | - | 3, 4 | 515 | |||
AK128195.1 | - | 15, 16 | 1869 | |||
BC159100.1 | - | 16, 17 | 2042 | 8860524 |