Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000507.3 | NP_000498.2 | 2 | 534 | ||
NM_001127628.1 | NP_001121100.1 | 3 | 433 | |||
XM_006717005.3 | XP_006717068.1 | 2 | 247 | |||
GenBank mRNA | AF073475.1 | - | 2 | 200 | ||
AK223395.1 | - | 2 | 478 | |||
AK292199.1 | - | 3 | 472 | |||
AK316583.1 | - | 2 | 535 | |||
AY544122.1 | - | 2 | 290 | |||
BC012927.1 | - | 2 | 400 | 4517633 | ||
D26054.1 | - | 2 | 209 | |||
D26055.1 | - | 2 | 209 | |||
D26056.1 | - | 2 | 209 | |||
L10320.1 | - | 2 | 345 | |||
M19922.1 | - | 2 | 401 |