Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021363.2 | NP_067338.2 | 2 | 383 | ||
GenBank mRNA | AF242220.1 | - | 2 | 368 | ||
AK020575.1 | - | 2 | 379 | |||
BC131996.1 | - | 2 | 383 | 40130273 | ||
BC131998.1 | - | 2 | 383 | 40130275 | ||
BY722061.1 | - | 2 | 379 |