Allergy to parrots has been reported to be associated with rhinoconjunctivitis, asthma, allergic rhinitis, bird-egg syndrome, and bird fancier’s lung (11-15).
Asthma, allergic rhinitis, and rhinoconjunctivitis
Bronchial asthma, allergic rhinitis, and rhinoconjunctivitis may result from exposure to budgerigar feathers, serum, or droppings (11, 15).
Bird-egg syndrome
Bird-egg syndrome (typically based upon sensitization to bird allergens and allergic symptoms occurring after egg consumption) has been reported in adults due to sensitization to avian airborne allergens through feathers, sera, and droppings of pet birds, including parrots (12).
Bird fancier’s lung
Bird fancier’s lung is a common type of hypersensitivity pneumonitis triggered by frequent exposure to avian aeroantigens (including proteins present on the feathers and excreted in bird droppings) (13, 14).
In a study involving patients with bird fancier’s asthma, the highest sIgE antibody binding was observed among bird fanciers (the bird caregivers) who had contact with the feathers of the bird species, including parrots (10).