These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Inflammatory bowel disease (IBD) is characterized by destructive inflammation of the intestinal tract with ulcerative colitis and Crohn’s disease as the most common disorders. Both diseases are life-long conditions with periods of remission and relapse.1
The EliA™ IBD portfolio, comprising the EliA Calprotectin 2 and the EliA™ ASCA IgA and IgG tests (ASCA: anti-Saccharomyces cerevisiae antibodies), supports the identification and the differentiation of IBD.
The follow-up of a positive EliA Calprotectin 2 test result with EliA ASCA IgA and IgG tests and a p-ANCA indirect immunofluorescence assay (IFA) helps differentiate between Crohn’s disease and ulcerative colitis, potentially avoiding further investigations and providing guidance for endoscopic procedures.9,10
The high sensitivity (98-99%) of the EliA Calprotectin 2 test, together with a negative predictive value (NPV) of 98.4 percent, make the test an outstanding tool helping to rule out inflammatory bowel disease.7,8
The EliA™ gastrointestinal portfolio* enables you to help rule in or rule out IBD (and subsequently differentiate between Crohn’s disease and ulcerative colitis), IBS, celiac disease, non-celiac gluten sensitivity, and wheat allergy.
The EliA gastrointestinal portfolio can be measured from one sample in one run and on the same fully automated Phadia™ Laboratory Systems.
*EliA Calprotectin 2, EliA ASCA IgA and IgG, EliATM Celikey IgA and IgG, EliATM GliadinDP IgA and IgG tests, ImmunoCAP™ Wheat allergen, f4
To support the needs of our customers, the EliA™ Stool Extraction Kit plus extraction device (Art. No. 14-6665-01) for the EliA™ Calprotectin 2 test was developed to offer an optimized stool extraction procedure.
The EliA Stool Extraction Kit plus extraction device helps you to increase the overall efficiency in your laboratory.
Total process time |
|
---|---|
EliATM Stool Extraction Kit 2 |
EliA Stool Extraction Kit plus |
59 min 39 sec |
45 min 33 sec |
Reduction of 24% |
Total process time |
---|
Total hands-on time |
|
---|---|
EliATM Stool Extraction Kit 2 |
EliA Stool Extraction Kit plus |
44 min 39 sec |
35 min 33 sec |
Reduction of 20% |
Total hands-on time |
---|