Allergenic molecules
Table adapted from Allergome.org (12).
Allergen
|
Source
|
---|
Cof a [Leaf]
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a [Seed]
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a 1
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a 1.0101
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a 2
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a 2.0101
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a 3
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
Cof a 3.0101
|
Coffea arabica, Coffee, Plants, Rubiaceae
|
The first allergen isolated from Coffea arabica was a class III chitinase with a molecular weight of 32 kDa (Cof a 1), which was identified in 3 out of 17 symptomatic coffee workers (18%) (13). Cof a 2 and Cof a 3 (9 and 7 kDa, respectively) are two cysteine-rich metallothioneins that have also been identified as coffee allergens. In a screening of 18 symptomatic coffee workers 8 (44%) had serum IgE antibodies to one of the two allergens (7, 14).
Cross-reactivity
One study examined the cross-reactivity between extracts of various coffee beans and dust as well as with castor bean, which is another suspected allergen in the coffee processing industry. However no evidence of cross-reactivity was found with the roasted coffee beans (15).