Despite its frequent consumption, allergic reactions due to onion ingestion are sporadic and limited to a small number of cases. Onions can induce both immediate and delayed hypersensitivity reactions. In sensitized individuals, onion handling can induce rhinoconjunctivitis, asthma and contact dermatitis (2).
One case study identified a 35-year-old man who had a severe systemic reaction/anaphylaxis requiring hospital treatment after ingesting cooked onions (2). Anaphylaxis was also reported in a 44-year-old patient after eating raw or lightly-cooked onions, with symptoms including intense itching, urticaria, confusion, blurred vision, transient loss of consciousness, sweating and tachycardia (8). Another patient, with a history of rhinoconjunctivitis and contact urticaria with mugwort and oral itching with peach skin, reported urticaria and oral itching after ingestion of raw onion (6).
Onions release irritating substances (thiopropanal sulfoxide and diallyl disulfide) when cut, which can cause eye irritation, respiratory and cutaneous allergic reactions. As a result, onions, along with the rest of the Amaryllidaceae family, have been implicated as a cause of occupational rhinoconjunctivitis and bronchial asthma (6).