Nature
Meadow fescue is a perennial grass with a height of 50 to 120 cm, with an erect branched panicle or nodding (10-20 cm long). Leaf blades are wide (4-8 mm) and flat. Spikelets are lance shaped, with a length of 8-12 mm. The pollen grains of Meadow fescue are spherical to ovoid in shape with a diameter of 31-40 µm (1).
Habitat
Meadow fescue is found in temperate regions, native to northern Europe and the United States. Ideally, it grows in open and damp places e.g. roadsides and waste places. It is a valuable pasture grass, but currently less planted (1).
Taxonomy
Timothy grass belongs to the phylum Spermatophyta, class Monocotyledonae, and subfamily Pooidaea (2). The genus Phelum consists of 15 species, among them P. pratense is the most extensively cultivated species (18). The two commonly recognized varieties are: i) P. Pratense var. pretense, ii) P. Pratense var. nodosum (18).
Taxonomic of Meadow fescue (1) |
---|
Domain |
Eukaryota |
Kingdom |
Plantae |
Family |
Poaceae |
Subfamily |
Pooidaea |
Genus |
Festuca |
Species |
Festuca pratensis |
Taxonomic of Meadow fescue (1) |
---|
Tissue
Pollen size: 36 (32.3-38.4) μm (medium).
Pollen class: 08 Monoporatae, 8.2. Poaceae.
Pollen grains are monoporate, spheroidal to ovoid, diameter of 31-40 mm, circular germinal pore of 2.5-4.5 mm diameter with a wavy margin, found at the larger end. Operculum has a diameter of 1.5- 2.0 mm with an irregular outline. The surface structure is fine but markedly granular (1).