Whole Allergen
i5 Yellow hornet
i5 Yellow hornet Scientific Information
Type:
Whole Allergen
Display Name:
Yellow hornet
Route of Exposure:
Sting
Family:
Vespidae
Species:
Dolichovespula arenaria (Vespula arenaria)
Latin Name:
Dolichovespula arenaria
Other Names:
Common yellow hornet, Aerial yellow jacket, Sandhills hornet
Summary
Yellow hornets (aerial yellow jackets) are members of the Vespidae (wasp) family found in temperate regions of Europe, Asia and North and South America. Vespid stings are common and cause mild to severe, localized and systemic allergic reactions including anaphylaxis, which may be fatal. Antigens 5 are the most abundant and allergenic proteins in vespid venom, demonstrating cross-reactivity between wasps, hornets and yellow jackets and also with fire ant venom and possibly mosquito and horsefly bites. Serological patterns of cross-reactivity are diverse and not necessarily correlated with clinical symptoms. There is conflicting evidence for cross-reactivity with bee venom.
Allergen
Nature
Yellow hornets (aerial yellow jackets) are members of the wasp family found in temperate regions of Asia, Europe and North and South America (1, 2). They are social wasps, living in large colonies and building large nests near human dwellings, often hanging from tree branches or the eaves of houses (2-4). Yellow hornets are not usually attracted by food and are not often in contact with humans; in general, they only sting when their nests are disturbed (2). It is difficult to distinguish between the appearance of hornets, paper wasps and yellow jackets (5).
Hymenoptera (bee, wasp and hornet) venoms contain a complex mixture of low-molecular-weight substances (biogenic amines, basic peptides, toxins) together with higher molecular weight proteins, including enzymes, and a variety of other components. All of these may contribute to sensitization, symptoms of allergy and the success of immunotherapy (5).
Taxonomy   
Taxonomic tree of Dolichovespula arenaria (6)
Domain
Eukaryota
Kingdom
Animalia
Phylum
Arthropoda
Subphylum
Hexapoda
Class
Insecta
Family
Vespidae
Genus
Dolichovespula
Taxonomic tree of Dolichovespula arenaria (6)
The Vespidae (wasp) family belongs to the order Hymenoptera, a large group of insects which includes bees, wasps, yellowjackets, hornets and ants (4).
Tissue
Venom.
Epidemiology
Worldwide distribution 
The exact incidence of Hymenoptera stings is unknown, although an estimated 56–94% of adults have been stung at least once during their lifetime. In a North American study of 3,236 Hymenoptera-allergic subjects, each person averaged 2.7 stings during their lifetime and 89% reported at least one systemic allergic reaction (4). The prevalence of severe allergic reactions to Hymenoptera venom is estimated at between 3% and 8.9% of adults (4, 7). Insect sting-related anaphylaxis accounts for over 30% of all anaphylaxis cases seen in emergency departments and almost 20% of all deaths from anaphylaxis (4). Sensitization to vespid venom is found in 10–30% of people with no history of allergic reaction to vespid stings (5).
Risk factors 
Large local reactions to Hymenoptera venom, defined as non-life-threatening pain, swelling, and erythema which may involve an entire limb, occur in 2.4% to 26.4% of the general population, but nearly 40% in people who are stung regularly, such as beekeepers (4). Other risk factors include a history of atopy, previous severe immunologic reactions, mast cell disorders, elevated serum tryptase and antihypertensive therapy with angiotensin-converting enzyme inhibitors (8)
Pediatric issues 
Severe allergic reactions to Hymenoptera venom are seen in up to 1% of children (4, 7).
Environmental Characteristics
Hymenoptera species play an important role in crop pollination and in reduction of insect pest populations (4).
Route of Exposure
Main 
The main route of exposure is a yellow hornet sting.
Clinical Relevance
Allergic reactions to hornet or yellow jacket stings are common; these are IgE-mediated and may be immediate or delayed. Most sting reactions are localized and self-limiting. Some present as a large local reaction: an area of swelling extending around the sting site and sometimes involving an entire limb or side of the face. These can be treated with ice packs, elevation and oral antihistamines and/or corticosteroids (2, 4).
Hymenoptera stings are one of the most frequent causes of severe anaphylaxis in adults. Symptoms include flushing, urticaria and angioedema as well as more serious gastrointestinal, respiratory and cardiovascular symptoms and multi-organ failure. Epinephrine (adrenaline) is the emergency treatment of choice (1, 4).
Around 10% of adults who have a mild systemic reaction to a sting will have a more severe reaction when stung again. Conversely, approximately 45% of patients with a moderate or severe reaction to their first sting will have a less severe reaction to future stings (3).
Almost 8% of subjects presenting with Hymenoptera allergy have mast cell disorders such as cutaneous mastocytosis. Patients with mast cell disorders and/or elevated baseline serum tryptase are at risk for more severe allergic reactions to vespid stings. Life-threatening sting reactions may be the only clinical presentation of these disorders (4, 5).
Anti-hypertensive therapy with angiotensin-converting enzyme inhibitors (ACEIs) and angiotensin receptor blockers (ARBs) may be associated with severe allergic reactions (including anaphylaxis) following Hymenoptera stings and/or immunotherapy for Hymenoptera venom allergies. These reactions are attributed to systemic accumulation of bradykinin (8).
Diagnostics Sensitization
Venom-specific IgE is most easily detected 1–4 weeks after the sting has occurred. A combination of serum IgE evaluation followed by intra-dermal skin prick testing is recommended for accurate diagnosis of sensitivity reactions; although the two tests show discordance, they are complementary (5, 9). There is no correlation between the severity of sting reactions and skin test reactivity to whole venom extracts (5).
Venom-allergic patients with concurrent mastocytosis are more likely to have negative specific IgE (sIgE) levels than venom-allergic patients without mastocytosis (5).
Recombinant allergenic proteins are preferable to native Hymenoptera venom when differentiating between bee and wasp venom sensitivity, because recombinant allergens do not contain cross-reactive carbohydrate epitopes (10).
Prevention and Therapy
Allergen immunotherapy
Dolichovespula arenaria is available as a single venom protein extract for venom immunotherapy (VIT). VIT is recommended where patients are IgE-sensitized to the culprit species and their systemic reaction is more severe than cutaneous involvement (3). VIT protects 75%–95% of patients from a systemic allergic reaction following a subsequent sting, compared to a 40%–60% risk of systemic reaction in untreated subjects. When systemic allergic reactions do occur following VIT, they are usually mild and not life-threatening. VIT should continue for a minimum of 3–5 years, although some patients require life-long therapy (4).
In children, VIT for systemic allergic reactions to Hymenoptera venom is safe and appears to be more effective in inducing long-term tolerance than in adults (4).
Prevention strategies 
Self-injectable epinephrine (adrenaline) should be considered for all patients with a history of a sensitivity reaction to vespid stings, particularly those with previous moderate to severe reactions or increased risk of exposure (4, 5).
Molecular Aspects
Allergenic molecules
Allergens of Dolichovespula arenaria (4, 5, 11).
Name
Type
Mass (kDa)
Dol a 1
Phospholipase A1
-
Dol a 2
Hyaluronidase
-
Dol a 5
Antigen 5
23
Name
Type
Mass (kDa)
Antigen 5 is the most abundant and allergenic protein in vespid (wasp and hornet) venom; they are a major group within the CAP (cysteine-rich secretory proteins, antigen 5 and pathogenesis-related 1 proteins) superfamily. They are mainly found in stinging and blood-feeding insects, although their function is unknown (1, 12). Antigen 5 sIgE reactivity patterns are very diverse. Some patients only react to antigen 5 and others react to all the allergenic protein groups. Some only react to antigen 5 from one vespid species. while others react to antigens 5 from various species in combinations that are not connected to the degree of antigen 5 structural homology (12). According to the WHO/IUIS allergen nomenclature database, the allergen sensitization rate for Dolichovespula arenaria antigen 5 is 81% (1).
Cross-reactivity
Vespid venoms are highly cross-reactive, especially among Vespula (wasp, yellow jacket), Dolichovespula (yellow or white-faced hornet, aerial yellow jacket) and Vespa (hornet) species (4). Antigens 5 show extensive cross-reactivity; this is more pronounced when using native venom extracts than with recombinant antigens 5 (12).
sIgE antibodies can be positive to both the native wasp and the native bee venom in up to 60% of patients, even though less than 10% of patients allergic to Hymenoptera venom demonstrate a systemic reaction to both wasp and bee stings. This double positivity may be due to cross-reactive carbohydrate determinants (CCDs) in both venoms, or the presence of sequence homology between proteins such as hyaluronidase or dipeptidylpeptidase (10, 12). However, some authors suggest that Vespidae (wasp and hornet) and Apidae (bee) venoms do not cross-react (4).
There is some evidence for cross-reactivity between the phospholipase A1 and antigen 5 components of Vespidae and fire ants (Solenopsis species) (3, 7). Wasp venom-allergic patients may also experience systemic reactions to the bites of mosquitoes or horseflies (1).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed:March 2022
References
  1. Blank S, Bazon ML, Grosch J, Schmidt-Weber CB, Brochetto-Braga MR, Bilò MB, et al. Antigen 5 Allergens of Hymenoptera Venoms and Their Role in Diagnosis and Therapy of Venom Allergy. Curr Allergy Asthma Rep. 2020;20(10):58.
  2. LOINC. Yellow Hornet IgE Ab/IgE total in Serum 2006 [cited 2022 11.01.22]. Available from: https://loinc.org/39544-2/%203/3%20https:/fhir.loinc.org/CodeSystem/$lookup/?system=http://loinc.org&code=39544-2].
  3. Korošec P, Jakob T, Harb H, Heddle R, Karabus S, de Lima Zollner R, et al. Worldwide perspectives on venom allergy. The World Allergy Organization journal. 2019;12(10):100067-.
  4. Pesek RD, Lockey RF. Management of Insect Sting Hypersensitivity: An Update. Allergy Asthma Immunol Res. 2013;5(3):129-37.
  5. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  6. ITIS. Dolichovespula arenaria  (Fabricius) 2021 [cited 2022 11.01.22]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=154259#null.
  7. Srisong H, Daduang S, Lopata AL. Current advances in ant venom proteins causing hypersensitivity reactions in the Asia-Pacific region. Mol Immunol. 2016;69:24-32.
  8. Diaz JH. Hypersensitivity Reaction to Yellow Jacket Sting in a Previously Nonallergic Patient Taking an Angiotensin Receptor Blocker. Wilderness Environ Med. 2017;28(2):150-2.
  9. Park HJ, Brooks DI, Chavarria CS, Wu RL, Mikita CP, Beakes DE. Combining Discordant Serum IgE and Skin Testing Improves Diagnostic and Therapeutic Accuracy for Hymenoptera Venom Hypersensitivity Immunotherapy. J Allergy Clin Immunol Pract. 2021.
  10. Šelb J, Kogovšek R, Šilar M, Košnik M, Korošec P. Improved recombinant Api m 1- and Ves v 5-based IgE testing to dissect bee and yellow jacket allergy and their correlation with the severity of the sting reaction. Clin Exp Allergy. 2016;46(4):621-30.
  11. allergome.org. Dolichovespula arenaria 2021 [cited 2022 11.01.22]. Available from: https://www.allergome.org/script/search_step2.php.
  12. Schiener M, Eberlein B, Moreno-Aguilar C, Pietsch G, Serrano P, McIntyre M, et al. Application of recombinant antigen 5 allergens from seven allergy-relevant Hymenoptera species in diagnostics. Allergy. 2017;72(1):98-108.