Whole Allergen
t15 White Ash Pollen
t15 White Ash Pollen Scientific Information
Type:
Whole Allergen
Display Name:
White Ash Pollen
Route of Exposure:
Inhalation
Family:
Oleaceae
Species:
Fraxinus americana
Latin Name:
Fraxinus americana
Other Names:
Fraxinus biltmoreana Beadle
Summary
White ash is a wind-pollinated tree, belonging to the genus Fraxinus of the family Oleaceae. The trees can grow at varied altitudes i.e., from plains to mountains, and can survive both, in low light and water levels. Fraxinus Americana (White ash) is widely distributed across Europe and North America and expanded to China, Pakistan, India, Afghanistan, Morocco, and Algeria. According to some data, where a large patient population (n=1,006) was taken, the prevalence of allergy to white ash was reported to be ~40%. Further, inhalation has been found to be the primary route of exposure to white ash pollen allergen. Ash pollen allergy can be prevented by avoiding stepping out and playing outdoor activities during peak hours and peak months of pollination It is one of the most prevalent allergens leading to asthma, allergic rhinitis, rhino-conjunctivitis, and atopic dermatitis. Closely related to olive, lilac, and privet, due to similar glycoproteins and high sequence identity, with limited cross-reactivity observed with tree, grass and weed pollens.
Allergen
Nature
White ash (F. americana) is a wind-pollinated tree that belongs to the family Oleaceae (1). The tree is huge in size and grows up to 120 feet in height. It is a long-lived tree and survives for almost 200 years. The trunk of the tree is straight and extends into the upper crown. They have shallow to slightly deep roots. Leaves are tapered at both ends and usually have 7 leaflets, oval to lance-shaped, pinnately opposite, with lengths of 6.35 to 15.34 cm. The spread of winged fruit (Samara) is facilitated by wind and water, and are single as compared to the double fruit of maples (2). 
Habitat
White ash trees can grow at varied altitudes i.e., from plains to mountains. It can be found in forests, riverside groves, as well as meadows (3). The tree requires fertile, well-drained sandy and clay loam soils. Soils with high nitrogen and calcium content, with a pH ranging from 5 to 7.5 helps in promoting its growth. The tree can survive both in low light and water levels. The average temperature range for the growth of the tree ranges from -14 to 27° C (4).
Taxonomy
Taxonomic tree of White Ash  (5)  
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Subphylum Angiospermae
Class Dicotyledonae 
Order  Oleales
Family Oleaceae
Genus Fraxinus
Species Fraxinus americana
Taxonomic tree of White Ash  (5)  
Tissue
White ash contains prolate to spheroidal shaped pollen grains that have 3-6 colpi. Pollen grains have reticulated (net-like) exine, and the size of each grain ranges from 18-28 microns (6). 
Epidemiology
Worldwide distribution
White ash is a native plant of Europe that is widely distributed in the entire continent (7).
It can be found throughout the world, that includes France, North America, North East Asia, China, Northern regions of Pakistan, India, Afghanistan, Morocco, and Algeria (8).
In a study conducted between the years 1997 and 1999 in Central Europe, a total of 12812 patients with suspected inhalant allergy were included. Of these patients, a total of 5416 (42.3%) were found to be pollen sensitized. Among the pollen sensitized patients, 17.6% (n=920) of patients were sensitive to ash pollens. Furthermore, in these 920 ash pollen sensitized patients,  9.5% (n=87) were reported to be mono-sensitized, 47.6% (n=438) were oligo-sensitized, and 42.9% (n=395) were polysensitized individuals (9).
In a cross-sectional study conducted in Saudi Arabia, that included 209 patients with allergic clinical presentation, 1.9% of patients displayed positive serum immunoglobulin (sIgI) for white ash (10).
In another cross-sectional study conducted between October 2010 and February 2014, a total of 1,006 allergic patients, aged between 1 to 86 years were included in the study. Of these patients, ~40% were found to be allergic to white ash. Furthermore, ash was considered to be one of the most prevalent allergen in spring (15.5%), summer (46.1%), autumn (22.7%), winter (15.7%) (11).
A survey was conducted in 476 adult allergy patients in Germany, that reported the prevalence of ash allergy (found by prick test reactivity) to be higher in North-Rhine Westphalia (28%) when compared to the Bavaria region (19%) (12).
Risk factors
Ash pollination period varies according to geographical regions. Pollination season begins early in January in California and South-central states, in April in the rest of the United States, and in May in Canada (2).
In recent years, sensitization with ash pollens is perceived as a recurrent cause of spring pollinosis. It is more frequent in polysensitized patients. The impact of allergy due to ash pollens is high due to its overlapping pollination period (birch or grasses pollen) and also because it is not included in routine screening assay procedures for inhalant allergies (13, 14). The rise in pollen production also increases the number of cases of allergic individuals (15). 
Environmental Characteristics
Living environment
White ash trees grow at diverse altitudes i.e., from plains to rocky mountains. It can be found either in forests, riverside seeps, slopes, lakes or meadows. Ash tree forests are known as frasinete (Fraxinetum) and are found to be rare (3).
Worldwide distribution
White ash trees are native to North America but are present throughout the world. It can be found from Nova Scotia in the west to Minnesota in the east, and Texas in the south to Florida in the north. It is also cultivated in Hawaii (16). It is comprised of approximately 65 species and grows in temperate areas of the Northern hemisphere (2).
In Europe, it is found from Ireland to the Russian Federation and from Scandinavia to Spain, Italy, and the Balkans. In Romania, it is considered the principal member of the Oleaceae family (3).
Route of Exposure
Main
The main route of exposure is through airway inhalation. Pollens from white ash trees are wind-pollinated. These pollens are carriers of allergens, however, exposure to the pure allergen is rare, instead, it is exposed through the particles of allergen pollen (17).  Intact pollens cannot enter the lower respiratory tract; and thus, affect the nasal or nasopharyngeal mucous membrane. However, particles of pollens can reach the lower respiratory tract, mainly through respiration by mouth (17).
Inhalation and penetration of pollen allergen particles to the lower respiratory tract is a direct cause of symptoms of pollinosis (17, 18).
White ash dispersion takes place, both by insects as well as wind (ambophily) (19). 
Clinical Relevance
Allergic rhinitis
The most common allergic disease that has an adverse effect on patients’ quality of life is allergic rhinitis (AR). The prevalence and pattern of sensitization vary geographically (20).
A cross-sectional study was conducted (October 2010 and February 2014) in Iran that evaluated the prevalence of ash pollen allergy in patients with AR (n=699; aged 1-86 years). The study reported that 61.1% of patients with AR developed ash pollen allergy (11).
Similarly, in a study conducted in Europe that investigated the prevalence of sensitization to ash pollen allergens in 5416 allergic patients, 17.6% of patients were sensitized to ash pollens (9). In another study performed in Iran, 97% of atopic patients were reported to be sensitized to white ash pollens (20).
Asthma
In a cross-sectional study involving 163 patients (aged between 1 to 86 years) with asthma, the prevalence of allergy to ash pollen was found to be 48% in these patients (11).
In a retrospective, observational study, a total of 113 patients, received nasal provocation test with ash pollen. Of these patients, 58% were white ash allergic to clinical symptoms, whereas 42% were without clinical symptoms. Furthermore, 49% and 45% of these patients reported asthma, respectively (21).
Atopic Dermatitis
In a cross-sectional study that enrolled 40 patients with atopic dermatitis, the prevalence of ash pollen allergy was reported to be 44.4% in these patients (11). 
Other diseases
Ash pollen allergy is commonly not prioritized by allergists. In Europe, birch-pollen allergy usually causes symptoms of rhino-conjunctivitis, and since the pollination period of ash and birch overlap (April/May), it is usually assigned to birch pollen (21).
A cross-sectional study was conducted between October 2010 and February 2014 to study the prevalence of ash pollen allergy in patients with urticaria (n=115; age 1-86 years). Based on the results it was reported that 37.5% of patients with urticaria developed ash pollen allergy (11).
Other topics
In a retrospective study conducted between December 2009 and March 2017, a total of 113 patients underwent nasal provocation and skin prick testing to ash pollen. According to the results, the nasal provocation test found that 58% of 113 patients being sensitized to ash were allergic with clinical symptoms, whereas 42% of patients were only sensitized without showing any symptoms (21).
Prevention and Therapy
Allergen immunotherapy
As majority of the patients with ash pollen allergy exhibit polysensitization, identifying the right allergen that causes sensitization is extremely essential for choosing proper allergen immunotherapy (AIT) since molecular diagnosis changes the indication for AIT (22).
Prevention strategies
Avoidance
Ash pollen allergy mainly takes place due to wind pollination. One should avoid going out and play outdoor activities during peak hours and peak months of pollination (23). 
Molecular Aspects
Cross-reactivity
Close phylogenetic relationships among Oleaceae members such as olive, lilac, and privet lead to cross-reactivity. Their main allergens, Ole e 1 (88%), Syr v 1 (82%), and Lig v 1 (91%), belonging to Ole e 1–like protein family, are responsible for high amino acidic sequential similarity with ash pollen allergen (Fra e 1) (7, 24). It is often suggested ash pollen allergic symptoms might be due to sensitization to cross-reactive allergens from other related or non-related sources. Various in vitro studies have shown cross-reactivity between ash pollen and birch pollens (7).
A study was conducted among 40 ash pollen-allergic patients to determine the profile of allergen components in ash pollen. According to the results, allergic symptoms to the pollens of ash are a consequence of sensitization to cross-reactive allergens from tree, grass and weed species (1).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: December 2020
References
  1. Niederberger V, Purohit A, Oster JP, Spitzauer S, Valenta R, Pauli G. The allergen profile of ash (Fraxinus excelsior) pollen: cross-reactivity with allergens from various plant species. Clin Exp Allergy. 2002;32(6):933-41.
  2. Weber RW. On the Cover. Annals of Allergy, Asthma and Immunology. 2004;93(1):6.
  3. Berghi NO. Oleaceae pollen allergy and its cross-reactivity in the mediterranean area in the context of globalization. 2014.
  4. Robin-Abbott MJ, Pardo LH. How climatic conditions, site, and soil characteristics affect tree growth and critical loads of nitrogen for northeastern tree species. Gen Tech Rep NRS-172 Newtown Square, PA: US Department of Agriculture, Forest Service, Northern Research Station 143 p. 2017;172:1-143.
  5. CABI. Fraxinus americana (white ash) 2019 [cited 2020 29.10.2020]. Available from: https://www.cabi.org/isc/datasheet/24506#totaxonomicTree.
  6. Shew HW. Getting to Know Our Local Pollen: Ash. Oleacea – Fraxinus sp Ash Trees [Internet]. 2017 [cited 2020. Available from: https://alabamaallergy.com/2017/05/17/getting-to-know-our-local-pollen-ash-2/.
  7. Barderas R, Purohit A, Papanikolaou I, Rodríguez R, Pauli G, Villalba M. Cloning, expression, and clinical significance of the major allergen from ash pollen, Fra e 1. Journal of allergy and clinical immunology. 2005;115(2):351-7.
  8. Sarfraz I, Rasul A, Jabeen F, Younis T, Zahoor MK, Arshad M, et al. Fraxinus: A plant with versatile pharmacological and biological activities. Evidence-Based Complementary and Alternative Medicine. 2017;2017.
  9. Hemmer W, Focke M, Wantke F, Gotz M, Jarisch R, Jager S, et al. Ash (Fraxinus excelsior)-pollen allergy in central Europe: specific role of pollen panallergens and the major allergen of ash pollen, Fra e 1. Allergy. 2000;55(10):923-30.
  10. Al-Rabia MW, Al-Rabia MW. Sensitization pattern to food and inhalant allergens in allergic patients from Jeddah city: Saudi Arabia. Webmed Central Allergy. 2016;7(8).
  11. Oskouei YM, Hosseini RF, Ahanchian H, Jarahi L, Ariaee N, Azad FJ. Report of common aeroallergens among allergic patients in northeastern Iran. Iranian journal of otorhinolaryngology. 2017;29(91):89.
  12. Höflich C, Balakirski G, Hajdu Z, Baron JM, Kaiser L, Czaja K, et al. Potential health risk of allergenic pollen with climate change associated spreading capacity: ragweed and olive sensitization in two German federal states. International Journal of Hygiene and Environmental Health. 2016;219(3):252-60.
  13. Hrabina M, Purohit A, Oster JP, Papanikolaou I, Jain K, Pascal P, et al. Standardization of an ash (Fraxinus excelsior) pollen allergen extract. Int Arch Allergy Immunol. 2007;142(1):11-8.
  14. Poncet P, Senechal H, Clement G, Purohit A, Sutra JP, Desvaux FX, et al. Evaluation of ash pollen sensitization pattern using proteomic approach with individual sera from allergic patients. Allergy. 2010;65(5):571-80.
  15. Villalba M, Rodriguez R, Batanero E. The spectrum of olive pollen allergens. From structures to diagnosis and treatment. Methods. 2014;66(1):44-54.
  16. NRCS U. WHITE ASH Fraxinus americana L. Plant Symbol = FRAM2 National Plant Data Center & the Biota of North America Program 2003.
  17. D'Amato G, Cecchi L, Bonini S, Nunes C, Annesi-Maesano I, Behrendt H, et al. Allergenic pollen and pollen allergy in Europe. Allergy. 2007;62(9):976-90.
  18. AnnaPomés, SabinaWünschmann, D.Chapman M. Allergens. Encyclopedia of Immunobiology. 2016;5:281-9.
  19. Wallander E. Evolution of wind-pollination in Fraxinus (Oleaceae). PhD diss Göteborg University. 2001.
  20. Fereidouni M, Hossini RF, Azad FJ, Assarezadegan MA, Varasteh A. Skin prick test reactivity to common aeroallergens among allergic rhinitis patients in Iran. Allergologia et immunopathologia. 2009;37(2):73-9.
  21. Eder K, Gellrich D, Meßmer C, Canis M, Gröger M. Component resolved analysis of ash pollen allergy in Bavaria. Allergy, Asthma & Clinical Immunology. 2018;14(1):76.
  22. Migueres M, Davila I, Frati F, Azpeitia A, Jeanpetit Y, Lheritier-Barrand M, et al. Types of sensitization to aeroallergens: definitions, prevalences and impact on the diagnosis and treatment of allergic respiratory disease. Clin Transl Allergy. 2014;4:16.
  23. Chad Z. Allergies in children. Paediatr Child Health. 2001;6(8):555-66.
  24. Asam C, Hofer H, Wolf M, Aglas L, Wallner M. Tree pollen allergens-an update from a molecular perspective. Allergy. 2015;70(10):1201-11.