Whole Allergen
t22 Pecan Pollen
t22 Pecan Pollen Scientific Information
Type:
Whole Allergen
Display Name:
Pecan Pollen
Route of Exposure:
Inhalation
Family:
Juglandaceae
Species:
C. illinoinensis
Latin Name:
Carya illinoinensis
Other Names:
Hickory tree
Summary
Pecan or Hickory is a deciduous tree and belongs to the Juglandaceae family. It grows to a height of 180 m and needs sufficient rainfall or watering with a humid to dry climate for cultivation. It is wind-pollinated, with the flowering season starting in spring. Typically, its pollen size is around 50 µm. A significant correlation of pecan pollen count with the onset of allergic symptoms in sensitized patients has been observed in studies. Pecan pollen is allergenic and has been reported to induce allergic rhinitis, conjunctivitis, and asthma in allergic patients. No pecan allergens have been identified yet. Cross-reactivity has been reported between genus Carya and Juglans species. 
Allergen
Nature
Pecan (Carya illinoensis) belonging to the Juglandaceae family, which consist of two important genus Carya (Pecan/Hickory) and Juglans (Walnut). It is a deciduous tree originating from North America. This tree can grow up to 180 feet in height and 83 inches in width. It is the largest among the hickory plant. It is a monoecious plant with male and female reproductive organs in the same plant. Flowering generally begins in the spring season. The pollen grain is pale yellow with three pores. It is wind-pollinated and is cable of producing pollen in large quantities (1). It is a good machining wood and thus valuable for flooring, furniture and paneling (2). Oil from the nuts is an ingredient in processed foods, is used in the manufacture of cosmetics and soap, and is a drying agent in paints(3).
Habitat
Pecan tree generally grows on the lowest portions of the river bottomland. This plant is sensitive to insufficient drainage. It has a rainfall requirement of around 1000 to 1300 millimeters. The preferred climate for pecan includes humid to dry but slightly more rain than an arid region and mild to chilly winter season (4).
Taxonomy
Taxonomic tree of Pecan  (5)  
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Subphylum Angiospermae
Class Dicotyledonae 
Order Juglandales
Family Juglandaceae
Genus Carya
Species Carya illinoinensis
Taxonomic tree of Pecan  (5)  
Tissue
Pollen of Carya illinoinensis is tetraploid, with the typical pollen size of about 43 to 53 µm (6).
Epidemiology
Worldwide distribution
Pecan is considered a major cause of allergic rhinitis in Northern America(7).
In a study performed in the United States in 209 children to test sensitization rate in rhinitis, 27.8% of children showed positive skin prick test towards pecan (8). Another study conducted on 371 allergic patients in New York City to analyze the tree pollen sensitization rate, observed hypersensitivity for pecan (hickory) in 27.1% of these patients (9).
In an Israeli study of 395 participants comprising 78.2% of a rural community and those atopic to 1 or more allergens, symptoms were seen in 50.3% had symptoms. 11.6% showed positive skin prick test to pecan, and 76.1% were atopic to pecan (p< 0.005). Of the individuals atopic to pecan, clinical symptoms coinciding only with the Pecan pollen season was observed in 65.2% of the atopic patients, and an additional 10.9% had perennial symptoms. The measurement of Pecan tree pollen during this period, May, showed that the pollen grains comprised 70% of the total airborne grains  (7). Another study in Israel confirms the same moderate correlation of pecan pollen count with the onset of allergic symptoms in sensitized patients. Pecan pollen count was observed to be 95% of total pollen count when pecan flowering season peaks in mid-May. In pecan allergic patients (n=26), around 58% (n=15) were symptomatic, most patients suffered from bronchial symptoms, and a few patients reported nasal or ocular symptoms(10).
In a study in Mexico, 4.8% of allergic patients (total n =247)  were found sensitive to pecan pollen (11). 
Environmental Characteristics
Worldwide distribution
Pecan is used in agricultural commodities in some parts of Mexico and the US (12). Pecan is also found in countries like Australia, Brazil, Israel, and South Africa (1).
Route of Exposure
Main
The main route of exposure for pecan pollen is through the airway (7).
Clinical Relevance
Pecan tree pollen is considered highly allergenic, resulting in allergic rhinitis, conjunctivitis and asthma in pecan sensitized individuals(7). The clinical symptoms such as nasal and bronchial onset were observed to be induced in pecan pollen sensitized individuals at an average concentration of 10-20 pollen per m3(10).
Allergic Rhinitis (AR) and Allergic Rhino-conjunctivitis (ARC)
A study was conducted in Israel to analyze the prevalence of pecan pollen allergy among 395 individuals. 11.6% were skin-prick test positive to Pecan tree pollen, constituting 25.4% of the atopic population. The part of individuals atopic to Pecan pollen was 76.1%, whereas 58.7% of the Pecan-atopic participants had AR (hay fever), 43.5% had asthma, and 31.5% had both AR and asthma. (7).
Asthma
Importance should be given to analyze the manifestation of pecan allergy and its symptoms in atopic children population (aged 4-17 years) as asthma prevalence has been found significant among pecan atopic individuals in such young population(7).
Other diseases
Walnut trees, especially black walnut (Juglans nigra) trees produce a highly toxic chemical called juglone. In the walnut tissue, juglone is present in non-toxic and colorless form hydrojuglone. When exposed to air or soil compounds, hydrojuglone is oxidized into juglone. Hydrojuglone is stored in leaves, stems, fruit hulls, inner bark, and roots. It is also used as an additive for food, preservative for wood and wood products, and an anti-bacterial or anti-fungal agent. Minute quantities can sicken, sedate, or kill living beings like other plants, animals, and humans(13).
Prevention and Therapy
Prevention strategies
Avoidance
Preventive measures such as taking a shower after coming home, removing clothes used at work outside the home, and abstaining from going outside when the pollen count is high, closing windows at home can help prevent exposure to pecan pollen (14). 
Molecular Aspects
Allergenic molecules
To date, no pecan tree pollen allergens have been identified. Though pecan food allergens, Car i 1, Car i 2 and Car i 4 have been identified and listed in the WHO/IUIS database(15). 
Cross-reactivity
Cross-reactivity could be expected between the genus Carya species and, to a moderate degree, to those of the genus Juglans, e.g., Walnut tree. Moderate to severe cross-reactivity has been demonstrated between shagbark hickory, pecan and black walnut(16, 17). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: November 2020
References
  1. Wetzstein HY, Rodriguez APM, Burns JA, Magner HN. Carya illinoensis (Pecan). In: Bajaj YPS, editor. Trees IV. Berlin, Heidelberg: Springer Berlin Heidelberg; 1996. p. 50-75.
  2. Peterson J. Carya illinoensis (Wangenh.) K. Koch pecan. Silvics of North America. 1990;2(1):205-10.
  3. Wickens GE, Chomsky N. Edible nuts: Food and Agriculture Organization of the United Nations Rome, Italy; 1995.
  4. Sparks D. Adaptability of Pecan as a Species. HortScience: a publication of the American Society for Horticultural Science. 2005;40.
  5. CABI. Invasive Species Compendium 2019 [22-11-2020]. Available from: https://www.cabi.org/isc/datasheet/17105.
  6. Stone DE. Pollen Size in Hickories (Carya). Brittonia. 1963;15(3):208-14.
  7. Rachmihl M, Verleger H, Waisel Y, Keynan N, Kivity S, Katz Y. The importance of the pecan tree pollen in allergic manifestations. Clinical & Experimental Allergy. 1996;26(3):323-9.
  8. Calabria CW, Dice J. Aeroallergen sensitization rates in military children with rhinitis symptoms. Ann Allergy Asthma Immunol. 2007;99(2):161-9.
  9. Lin RY, Clauss AE, Bennett ES. Hypersensitivity to common tree pollens in New York City patients. Allergy Asthma Proc. 2002;23(4):253-8.
  10. Waisel Y, Mienis Z, Kosman E, Geller-Bernstein C. The partial contribution of specific airborne pollen to pollen induced allergy. Aerobiologia. 2004;20(4):197-208.
  11. Ramos Morín CJ, Canseco González C. [Hypersensitivity to common allergens in the central region of Coahuila]. Rev Alerg Mex. 1993;40(6):150-4.
  12. Gallegos‐Infante JA, Rocha‐Guzman, N.E., Gonzalez‐Laredo, R.F. and Moreno‐Jimenez, M.R. Pecans ( Carya illinoinensis ).  Fruit and Vegetable Phytochemicals2017. p. 1137-44.
  13. Coder KD. Black Walnut Allelopathy: Tree Chemical Warfare. Warnell School of Forestry & Natural Resources, University of Georgia. 2017.
  14. ThermoFisher. Allergy insider: Thermo Fisher Scientific Inc.; 2018 [23-11-2020]. Available from: https://www.thermofisher.com/diagnostic-education/patient/in/en/allergy-resources/allergens/beech-tree.html.
  15. WHO/IUIS. Pecan food allergens 2019 [23-11-2020]. Available from: http://www.allergen.org/search.php?allergensource=carya&searchsource=Search.
  16. Weber RW. Cross-reactivity of pollen allergens: impact on allergen immunotherapy. Ann Allergy Asthma Immunol. 2007;99(3):203-11; quiz 12-3, 31.
  17. Yman L. Botanical relations and immunological cross-reactions in pollen allergy: Pharmacia diagnostics; 1983.