Whole Allergen
e206 Rabbit serum proteins
e206 Rabbit serum proteins Scientific Information
Type:
Whole Allergen
Display Name:
Rabbit serum proteins
Route of Exposure:
Inhalation
Family:
Leporidae
Species:
Oryctolagus cuniculus
Latin Name:
Oryctolagus cuniculus
Other Names:
Domestic rabbit, European rabbit, Rabbit (1)
Summary
Serum albumins of pets, including Oryctolagus cuniculus (rabbit), are inhalant allergens that cause respiratory symptoms in sensitized individuals. As rabbits are the third most common household pet in Europe and the US, rabbit allergens are an important cause of allergic respiratory disease.
Allergen
Nature
The dander and saliva of rabbits contain serum albumins that are involved in IgE-mediated cross reactions (2). Sensitization to serum albumins can occur by inhalation as well as ingestion, resulting in allergic respiratory symptoms (2) such as rhinitis, conjunctivitis, asthma and, on rare occasions, anaphylaxis (3)
Taxonomy 
Taxonomic tree of Oryctolagus cuniculus (1)
Domain
Eukaryota
Kingdom
Metazoa
Phylum
Chordata
Subphylum
Vertebrata
Class
Mammalia
Order
Lagomorpha
Family
Leporidae
Genus
Oryctolagus
Taxonomic tree of Oryctolagus cuniculus (1)
Tissue 
Dander and saliva (2).
Epidemiology
Worldwide distribution 
In patients with pet allergies, serum albumins are recognized by the serum of 20–30% of patients (Diaz-Perales et al, 2013). In a group of patients sensitized to rabbit allergens, IgE prevalence to Ory c 3 was 77% (4).
Risk factors 
Contact with rabbits in a professional setting, for example an animal testing laboratory, increases the risk of occupational sensitization. Risk factors for the development of laboratory animal allergies include atopy, familial history of allergic diseases, and environmental susceptibility (3).
As rabbits are popular pets, exposure to rabbit allergens occurs in the home through rabbit ownership or occasional contact (5). In addition to direct contact with rabbits, airborne particles from rabbit dander may settle then be re-dispersed into the air, causing allergic symptoms even when the animal is not present (2).
Environmental Characteristics
Worldwide distribution 
In Europe and the US, 30–60% of all households have one or more pets; rabbits are the third most popular pet in Europe and the US after cats and dogs (4).
Route of Exposure
Main
Inhalation (2)
Secondary
Ingestion (2)
Clinical Relevance
Symptoms of sensitization to rabbit allergens include allergic rhinitis, conjunctivitis, bronchial asthma and, on rare occasions, anaphylaxis (3).
Molecular Aspects
Allergenic molecules
Ory c 1, a 17-kDa glycoprotein found in saliva and fur, is a major rabbit allergen that can bind both IgE and IgG from serum of sensitized patients. In the same study, the authors suggest that rabbit serum albumin (Ory c 6) is a minor allergen (3). Ory c 1 and Ory c 4 (both lipocalins) are homologues with the cow allergen Bos d 5; additionally, Ory c 4 showed amino acid identity ranging between 24-63% homology with other domestic animal lipocalins (6).The table below, adapted from Allergome.org (7), highlights the rabbit serum, dander and saliva allergens.
Allergen
Type
Mass (kDa)
Ory c 1 [saliva, dander]
Lipocalin, Odorant binding protein
1.8
Ory c 3 [dander]
Lipophilin
22.5
Ory c 4 [saliva]
Lipocalin
19.7
Ory c 6 [serum]
Albumin
69
Ory c Phosvitin [dander, saliva]
Casein kinase
25
Allergen
Type
Mass (kDa)
Cross-reactivity
Ory c 4 has a high sequence identity to Fel d 4 and Can f 6, which makes IgE-cross reactivity highly probable (4).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed:March 2022
References
  1. CABI. Oryctolagus cuniculus (rabbits) 2021. Available from: https://www.cabi.org/isc/datasheet/72046.
  2. Díaz-Perales A, González-de-Olano D, Pérez-Gordo M, Pastor-Vargas C. Allergy to uncommon pets: new allergies but the same allergens. Front Immunol. 2013;4:492-.
  3. Choi JH, Kim HM, Park HS. Allergic asthma and rhinitis caused by household rabbit exposure: identification of serum-specific IgE and its allergens. J Korean Med Sci. 2007;22(5):820-4.
  4. Curin M, Hilger C. Allergy to pets and new allergies to uncommon pets. Allergol Select. 2017;1(2):214-21.
  5. Liccardi G, Piccolo A, Dente B, Salzillo A, Noschese P, Gilder JA, et al. Rabbit allergens: a significant risk for allergic sensitization in subjects without occupational exposure. Respir Med. 2007;101(2):333-9.
  6. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  7. allergome.org. Rabbit 2021 [cited 2022 19.01.22]. Available from: http://www.allergome.org/script/search_step2.php.