Whole Allergen
f1 Egg white
f1 Egg white Scientific Information
Type:
Whole Allergen
Display Name:
Egg white
Route of Exposure:
Ingestion
Family:
Phasianidae
Species:
Gallus gallus
Latin Name:
Gallus spp.
Other Names:
Egg white
Summary
Hen’s egg (Gallus gallus domesticus) is a protein-rich food source and is usually consumed as raw, cooked, or processed. Egg white is rich in proteins and is regarded as highly allergenic. The egg has been considered one of the most common food allergy sources, particularly in infants and children. Around 0.5 – 2.5 % of young children within a normal population have been found to be allergic to eggs. However, the prevalence of egg allergy in adults is less than 0.25 %. Egg sensitization, having IgE antibodies to egg, is a prerequisite for egg allergy. Egg sensitization is more prevalent than egg allergy, and the prevalence rate has been reported to be up to 6 % depending on age. The specific IgE (sIgE) test for egg white is commonly used when investigating a patient with a suspicion of egg allergy. Most egg-allergic children can develop tolerance to egg proteins over time. A patient's highest recorded egg white sIgE is significantly related to the persistence of egg allergy. A higher egg white sIgE value denotes a more extended period to develop tolerance. Egg IgE is predictive of allergy outcome and should be used in counseling patients on prognosis.
Egg white consumption may lead to allergic symptoms, such as gastrointestinal reactions, respiratory symptoms, cutaneous reactions, and anaphylaxis among egg white allergic patients. Therefore, egg-allergic individuals are usually recommended to avoid egg and egg-based food products strictly.
Moreover, egg allergy children are often falsely denied influenza or MMR vaccination as the vaccine might contain a small amount of egg protein. However, several studies and guidelines have claimed these vaccines to be safe for children with egg allergies.
An oral food challenge is considered to be the gold standard for diagnosing egg allergy. Egg sIgE testing can aid in diagnosing egg allergy, indicate when an egg challenge is safe to perform, and is predictive of egg allergy outcome. This means that the egg white test could be utilized in counseling patients on prognosis. Furthermore, egg allergy or egg white sensitization has been found to elevate the risk of developing sensitization to aeroallergens (such as grass pollens, house dust mites, cats, and dogs).
Early introduction of egg into the infants’ diet has been found to be beneficial in developing tolerance to the egg; it is presently studied as a means of primary prevention of egg allergy. Additionally, oral immunotherapy (for egg) is also performed in some countries on egg-allergic patients who fail to develop natural tolerance to the egg.
Allergens
Nature
Egg (Gallus gallus) is a food product with substantial nutritional content, suitable for all ages. Egg white is rich in proteins, mainly composed of water and lacks lipids (1). The egg is reported to contain both macro and micronutrients required for embryonic development (1).
Egg white of chicken (Gallus domesticus) constitutes the albumin portion present in the egg (2). It primarily contains water (approximately 88%) and more than 20 glycoproteins (like ovalbumin), fibrous structural proteins (ovomucins), antibacterial proteins (lysozyme), and peptides (1, 3). Egg white is regarded as the principal source of allergen; the egg white proteins are potentially more allergenic compared to protein derived from egg yolk (4-6). The major allergens found in egg white include ovomucoid (Gal d 1), ovalbumin (Gal d 2), ovotransferrin (Gal d 3), and lysozyme (Gal d 4) (4, 7). Globally, eggs have been extensively utilized in food products like bread, egg noodles, pancakes, waffles, cakes, French toast, tea-boiled egg, and others (8).
Nature
Hen’s eggs are regarded as fresh up to 28 days post laying. Room temperature or preferably refrigeration is considered ideal for the storage of shelled eggs. Variation in egg storage condition and duration can induce physiochemical changes and further protein denaturation. These changes are manifested due to the exchange of water between egg white and yolk and the loss of carbon dioxide and water through eggshells' pores. This further results in the rise of air cells between the two membranes of the eggshell. Moreover, with storage duration, the albumen height reduces, whereas the whipping volume and pH of albumen surge. (1).
Taxonomy
Gallus domesticus (hen’s egg) belongs to the family Phasianidae (chicken) (9). A study confirmed high cross-reactivity between hen’s egg white with egg white from turkey, duck, and goose belonging to the order Anseriformes (10). 
Taxonomic tree of Hen’s Egg (9)
Domain Eukaryota
Kingdom Animalia
Phylum Chordata
Subphylum Vertebtata
Class Aves
Order Galliformes
Family Phasianidae
Subfamily Phasianinae
Genus Gallus
Species Gallus gallus
Sub-species Gallus gallus domesticus
Taxonomic tree of Hen’s Egg (9)
Tissue
The average protein concentration of a whole egg is found to be 12.5g/100g of a raw fresh egg, while the average protein concentration of egg white (including vitelline membrane) might be 10.9g/100g of a raw fresh egg. The mean egg white volume has been estimated to be around 30ml (egg of 60-gram weight, including eggshell) (1). Egg white is primarily composed of protein (11%), water (88%), trace amounts of carbohydrates, ash, and lipids (1% in total). Further, the proteins present in egg white are categorized as major and minor proteins (11). The major protein comprises of ovalbumin (54%), ovotransferrin (12%),  ovomucoid (11%), and lysozyme (3.4%) (2, 12) .While, the minor proteins include ovoinhibitor (1.5%), ovoglycoprotein (1%), ovoflavoprotein (0.8%), ovomacroglobulin (0.5%), cystatin (0.05%) and avidin (0.05%) (11).
During the cooking process, the proteins in egg white have been found to usually lose their allergenicity due to conformational modifications (epitope destruction or epitope blocking) leading to protein denaturation (3, 13). This process of protein denaturation may lead to the inactivation of antinutritional factors like antiprotease from egg-white and other highly resistant proteins in eggs (1). 
Epidemiology
Worldwide distribution
The egg has been reported as one of the common inducers of food allergy prevalent in industrialized countries (14). According to a study, around 0.5-2.5% of young children have been observed to be affected with egg allergy (15, 16). A high prevalence of childhood egg allergy has been reported in Japan and France (4). According to a study conducted in Japan among 24 egg white allergic patients, 92% showed reactivity towards the egg white allergen (Gal d 1, ovomucoid) (17). An Italy-based study involving 104 suspected egg-allergic patients (0.7-15.1 years) reported 43.5% (20/46), 52.1% (24/46), and 36.9% (17/46) of patients to exhibit allergenicity towards Gal d 1, Gal d 2, and Gal d 4, respectively (18). Furthermore, a US-based cohort study also detected the prevalence of egg allergy to be approximately <0.25% (from graphical representation) in 4425 adults (20-60+ years) (19).
Sensitization to the egg white means that IgE antibodies are produced towards the egg white and is a prerequisite for an IgE-mediated egg allergy. Egg white sensitization is more prevalent than egg allergy, and IgE antibodies have been detected already in very young age groups. Tedner et al. found in a Nordic population-based birth-cohort study the prevalence of egg white sensitization among 3-month old infants to be 3.7% (41/1102)  (20). A German-based population study conducted on 14,836 children and adolescents (3-17 years) found the overall sensitization rate towards egg white was 5% (21). Moreover, a Japan-based cohort study found 0.8% of the children (at 9 years) to be sensitized towards egg white protein (Gal d 1) (22). A Taiwanese study on 2256 allergic children (0.1-17.6 years) detected the positive sensitization rate for egg white was highest  (53.5%) among children of age 2-4 years (12). Besides, egg white sensitization among the adult population has been reported in few studies. A European cohort study found an overall prevalence of egg white sensitization to be 0.4% among the adult (20-39 years) population (23). Furthermore, a US-based cohort study reported the prevalence of egg white sensitization in the adult population ranging between 2.1% to 3.9% among 4425 adults (20-60+ years) (19).
The natural history of egg white allergy
Food allergy during childhood could be a transitory condition, although it can be a persistent one in some cases. Hen’s egg has been reported to be one of the frequent food allergens affecting children during the first 10 years of life  (24). Egg allergy has been claimed to get resolved naturally with aging (2-6 years)  (5, 25). A study conducted on 226 egg allergic children (<6 years) reported tolerance development in 30% (66/226) of children by the age of 3 years, while in 73% (164/226) of children by 6 years (26). Savage et al. examined the relationship between the peak egg white IgE level and the development of tolerance while studying the natural history of egg allergy. The tolerance development rate was found to be inversely proportional to the peak level of egg IgE (P < 0.0001). The fastest rate of tolerance development was found in children with a peak egg IgE <2 kU/L, whereas tolerance development rate was slower in children with peak IgE between 2 and 49.9. Nonetheless, the majority of the children with the peak IgE ranging between 2 to 49.9 kU/L developed tolerance towards egg subsequently. The tolerance rate was slowest among children with peak egg IgE ≥50 kU/L. These children mostly developed tolerance, not before 18 years (the end of the study period). For instance, the tolerance development among the study population was found to be 46% (peak egg IgE <2 kU/L), 32% (peak IgE 2 to.4.9 kU/L), 17% (IgE peak level between 5 to 9.9 kU/L), 16% (peak level of 10 to 19.9 kU/L), 14% (peak IgE 20 to 49.9 kU/L), and 11% (peak sIgE level ≥50 kU/L) by 8 years of age. The authors concluded egg IgE to be a predictor of the outcome of allergy and, therefore, should be utilized in patients’ counselling on prognosis(27).
Moreover, according to some studies, the majority of the egg-allergic children showing reactions to lightly cooked regular egg preparations (scrambled eggs or French toast) were found to be tolerant towards baked egg products (muffin, waffle, cakes, etc.) (28-30). This tolerance towards baked egg products was indicative of the immunological shift towards the development of tolerance for regular eggs (28).  
Risk factors
Egg allergy is associated with atopic dermatitis (AD), asthma, rhinitis, and other food allergies (such as peanut) (2, 4, 12, 15). Clinical allergy towards eggs has been found to be associated with risk factors like male gender, young age, ethnicity/race (19), and family history (31). Moreover, the presence of specific IgE (sIgE) towards all four egg allergens (Gal d 1, 2, 3, or 5) has been reported to elevate the risk of developing persistent raw egg allergy (32).
Children with egg allergy are often denied influenza vaccination because the vaccine contains a small amount of egg protein. However, recent studies have demonstrated that children with even severe egg allergy can safely receive the vaccine. Therefore, based on this outcome, the current guidelines recommend routine immunization (influenza vaccination) of such egg-allergic patients without undergoing any testing or special considerations  (33). Moreover, as per the recommendations of vaccination-specific guidelines, the influenza vaccine can be received under usual clinical settings (34). A study conducted on egg-allergic patients immunized with live attenuated influenza vaccine reported no systemic reactions (allergic) or anaphylaxis among these patients post-immunization (35).
National Advisory Committee on Immunization (NACI) has deemed patients with egg allergy suitable for receiving the Measles Mumps Rubella (MMR) vaccination. According to this guideline, the immunization may be administered without prior testing but under proper guidance and adequate facility (36).
Pediatric issues
A significant correlation exists between sensitization towards outdoor/indoor aeroallergens (like grass pollens, house dust mites, cat, dog) and egg allergy during infancy (6, 37). Additionally, the coexistence of egg allergy and eczema at infancy has also been reported as a predictive indicator of aeroallergen sensitization and respiratory allergies among children (4 years) (37).
Environmental Characteristics
Worldwide distribution
Hen’s egg is consumed globally in various forms like raw, semi-cooked (poached or soft-boiled eggs) (1), completely cooked (hard-boiled, scrambled, baked, or fried) (38) or as an additive in products like bread, waffles, cakes and others (8). Additionally, eggs are also utilized in the cosmetic, pharmaceutical (especially vaccine preparation), and beverage refining (wine and beer) industry (38).
Route of Exposure
Main
The route of exposure is through consumption of egg-white(39). 
Clinical Relevance
Egg allergy has been claimed as one of the most frequent food allergies prevalent in children (6). Egg white consumption may lead to allergic symptoms, such as gastrointestinal (GI) reactions, respiratory symptoms, cutaneous reactions, and anaphylaxis in individuals allergic to egg white (40-42).
Anaphylaxis
An elevated egg white sIgE level has been reported to be correlated with anaphylaxis (43). As per a study, serum sIgE level towards egg white has been found to be higher (84.8%; 28/33) in patients with a history of anaphylaxis post-consumption of egg or egg derivatives (44). An OFC study on 374 egg white-allergic patients reported anaphylaxis among 2% (8/374) of the patients (40). Moreover, a Japanese study reported anaphylaxis in 8% (3/38) of the patients allergic to both heated and raw egg white (42).
Gastrointestinal (GI) reactions 
GI symptoms are mostly seen as immediate hypersensitivity reactions in egg-allergic individuals (6). As per an OFC study, 9% (34 out of 374) of egg white-allergic children exhibited GI reactions (40). In another OFC study, 84% (104/124) of patients (egg-sensitized; 1-19 years) showing positive response towards heated egg white challenge reported GI symptoms (41).
Skin reactions and Atopic dermatitis
Cutaneous reactions due to egg white allergy usually include urticaria, erythema, angioedema, and oral allergy syndrome (OAS). In an OFC study (with cooked and raw egg white), 95% (37/39) of children with positive challenge showed cutaneous reactions such as urticaria (28/37), erythema (4/37), OAS (4/37), and angioedema (2/37) (45). Another OFC study (with egg white) found cutaneous reactions in 98% (368/374) of egg-white allergic patients (40). Additionally, as per an OFC study, 41% (51/124) and 48% (59/124) of patients (egg-sensitized; 1-19 years) showing positive response towards heated egg white challenge reported skin symptoms and OAS, respectively (41).
Children with AD have been commonly found to show sensitization towards hen’s egg (41). A study reported AD in 87% (33/38) of the patients allergic to both heated and raw egg white (42).
Diagnostics Sensitization
Challenge test
Oral food challenge is considered the gold standard for diagnosing egg allergy. However, this diagnosis mode is often regarded as time extensive, resource-consuming, and potentially risky (5, 18, 44). 
In-vitro diagnostics
Egg white sIgE testing is used as an aid in diagnosing egg allergy and to monitor the disease development. The sIgE diagnostic values have been reported to be beneficial for physicians in deciding the necessity of an egg challenge while estimating its potential risk on the patients’ health. Furthermore, an age-specific correlation has been observed between egg sIgE levels and OFC outcome; younger children usually exhibit reactivity to even at low egg sIgE levels in contrast to the older children (40). Therefore, periodic monitoring of the absolute values of egg specific-IgE antibody level combined with a clinical history of egg allergy and physical examination can accurately help diagnose egg allergy as well as indicate when an egg challenge should be performed (2, 5, 13, 47). A prospective study conducted on 81 children (<2years; suspected of egg allergy) diagnosed (utilizing cooked and raw egg white) egg allergy among 79% (56/81) of these children, and egg white sIgE testing has been reported to show high diagnostic efficacy (45).   
Prevention and Therapy
Prevention strategies
Complete avoidance of egg and egg-based products from the diet is the recommended treatment for egg allergy (2, 39, 48); however, this could be challenging as egg is a common ingredient of many food and accidental intake of egg or egg-based products are usually quite common (5, 48).  
Patients allergic to heat-labile egg protein (like ovalbumin) might be tolerant towards cooked, boiled, or fried form of eggs (39). Hence, differential avoidance is typically recommended for egg-allergic patients based on their reactivity towards raw or cooked eggs.  Here, component resolved diagnosis (CRD) with egg components can be helpful in giving personalized avoidance advice. Moreover, reactivity towards raw or slightly cooked eggs has been found to be more prevalent compared to extensively processed egg-based food (42).
Early introduction of eggs into the diet has been found to be potentially beneficial in the prevention of egg allergy (14).  In a study conducted on 231 infants allergic to egg, early introduction of egg (4-6 months age) was found to be correlated with a reduced risk of egg allergy. In contrast, late introduction of an egg-based diet (at 10 months) might increase the risk of developing egg allergy (49). In general, guidelines recommend the necessity of more studies before early introduction of egg into the diet of an infant.
Allergen immunotherapy
Currently, allergen immunotherapy has been attempted to treat egg-allergic patients in addition to induction of oral tolerance (48). A study was conducted on 30 egg-allergic children (≥4 years) to estimate the efficacy of oral immunotherapy (OIT)-egg desensitization and evaluate the maintenance of tolerance. All the patients (n=16) attained desensitization towards egg after 4 months of therapy, while 31% were reported to retain tolerance towards egg even after 3 months of avoiding egg consumption  (50). 
Molecular Aspects
Allergenic molecules
Egg white possesses 23 distinct proteins and is considered a vital allergen source from egg (4, 7, 51).
However, only four allergenic proteins (listed below in the table) from egg-white of G. domesticus (i.e., Hen’s egg) have been officially recognized in the IUIS database (52). 
Allergen 
Biochemical Name 
Molecular Weight 
Allergenicity 
Gal d 1
Ovomucoid
28 kDa
·        Gal d 1 is a strongly allergenic, heat-resistant protein (53) [p169] and contributes 11% of protein content in egg white (54).
·       According to an OFC study, 43.5% (20/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 1 (18)
Gal d 2
Ovalbumin
44 kDa
·       Gal d 2 is heat-unstable and therefore bears the risk of manifesting clinical reactivity towards the raw or slightly heated egg (53) [p169]; it contributes 54% of protein content in egg white (54).
·       According to an OFC study, 52.1% (24/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 2 (18).
Gal d 3
Ovotransferrin
78 kDa
·       • Gal d 3 is thermo-labile and can manifest the risk of clinical reactivity towards raw or slightly heated egg (53)[p169]; it contributes 12% of the protein content in egg white (54).
·       According to an OFC study, 13.0% (6/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 3 (18).
Gal d 4
Lysozyme C
14 kDa
·       Gal d 4 contributes 3% of protein content in egg white (54).
·       According to an OFC study, 36.9% (17/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 4 (18).
Allergen 
Biochemical Name 
Molecular Weight 
Allergenicity 
IgE: Immunoglobulin E, kDa: kilodaltons, EA: Egg Allergy, ES: Egg-sensitized
Apart from the above-mentioned egg-white allergens, ovomucin (165 kDa; containing a trypsin inhibitor-like domain), a heavily glycosylated protein with strong antiviral activity (53) [p169] and ovoinhibitor (belonging to Kazal protease inhibitor family) are also reported to be present in egg white (55). 
Biomarkers of severity
As per a study, the best diagnostic biomarkers for raw and cooked egg allergy are distinct. Studies have found ovomucoid (Gal d 1) sIgE as a superior biomarker for cooked egg allergy  (51, 56-59). Additionally, ovalbumin (Gal d 2) is considered the best marker for diagnosing raw and cooked egg allergy (56). 
Cross-reactivity
A study reported cross-reactivity of the majority of allergens found in hen’s egg white with egg white proteins from turkey, duck, goose, and seagull (10). Moreover, egg white protein (Gal d 1) has been found to exhibit cross-reactivity with proteins present in egg yolk (Gal d 5 and Gal d 6) (57).
Furthermore, an immunoreactive allergen (66 kDa) isolated from the bird’s nest is claimed to be homologous with ovoinhibitor protein found in hen’s egg white, resulting in cross-reactivity between the two allergenic proteins (60). Additionally, livetin (allergenic protein) mediated cross-reactivity between hen’s egg proteins and bird dander has also been reported (61).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr Magnus Borres
Last reviewed: November  2021
References
  1. Rehault-Godbert S, Guyot N, Nys Y. The Golden Egg: Nutritional Value, Bioactivities, and Emerging Benefits for Human Health. Nutrients. 2019;11(3).
  2. Caubet JC, Kondo Y, Urisu A, Nowak-Wegrzyn A. Molecular diagnosis of egg allergy. Curr Opin Allergy Clin Immunol. 2011;11(3):210-5.
  3. Bartnikas LM, Sheehan WJ, Larabee KS, Petty C, Schneider LC, Phipatanakul W. Ovomucoid is not superior to egg white testing in predicting tolerance to baked egg. J Allergy Clin Immunol Pract. 2013;1(4):354-60.
  4. Benhamou AH, Caubet JC, Eigenmann PA, Nowak-Wegrzyn A, Marcos CP, Reche M, et al. State of the art and new horizons in the diagnosis and management of egg allergy. Allergy. 2010;65(3):283-9.
  5. Chokshi NY, Sicherer SH. Molecular diagnosis of egg allergy: an update. Expert Rev Mol Diagn. 2015;15(7):895-906.
  6. Caubet JC, Wang J. Current understanding of egg allergy. Pediatr Clin North Am. 2011;58(2):427-43, xi.
  7. Borres MP, Maruyama N, Sato S, Ebisawa M. Recent advances in component resolved diagnosis in food allergy. Allergol Int. 2016;65(4):378-87.
  8. Verhoeckx KCM, Vissers YM, Baumert JL, Faludi R, Feys M, Flanagan S, et al. Food processing and allergenicity. Food Chem Toxicol. 2015;80:223-40.
  9. Al-Nasser A, Al-Khalaifa H, Al-Saffar A, Khalil F, Al-Bahouh M, Raghbeb G, et al. Overview of chicken taxonomy and domestication. World's Poultry Science Journal. 2007;63.
  10. Langeland T. A clinical and immunological study of allergy to hen's egg white. VI. Occurrence of proteins cross-reacting with allergens in hen's egg white as studied in egg white from turkey, duck, goose, seagull, and in hen egg yolk, and hen and chicken sera and flesh. Allergy. 1983;38(6):399-412.
  11. Abeyrathne E, Lee H, Ahn D. Egg white proteins and their potential use in food processing or as nutraceutical and pharmaceutical agents—A review. Poultry Science. 2013;92(12):3292-9.
  12. Lin YT, Wu CT, Huang JL, Cheng JH, Yeh KW. Correlation of ovalbumin of egg white components with allergic diseases in children. J Microbiol Immunol Infect. 2016;49(1):112-8.
  13. Machinena A, Riggioni C, Dominguez O, Gereda D, Jimenez-Feijoo R, Folque M, et al. Are there any biomarkers that can predict tolerance to baked egg in egg allergic children younger than 6 years? Pediatr Allergy Immunol. 2020;31(4):427-30.
  14. De Martinis M, Sirufo MM, Suppa M, Ginaldi L. New Perspectives in Food Allergy. Int J Mol Sci. 2020;21(4).
  15. Sicherer SH, Wood RA, Vickery BP, Jones SM, Liu AH, Fleischer DM, et al. The natural history of egg allergy in an observational cohort. J Allergy Clin Immunol. 2014;133(2):492-9.
  16. Rona RJ, Keil T, Summers C, Gislason D, Zuidmeer L, Sodergren E, et al. The prevalence of food allergy: a meta-analysis. J Allergy Clin Immunol. 2007;120(3):638-46.
  17. Takagi K, Teshima R, Okunuki H, Itoh S, Kawasaki N, Kawanishi T, et al. Kinetic analysis of pepsin digestion of chicken egg white ovomucoid and allergenic potential of pepsin fragments. Int Arch Allergy Immunol. 2005;136(1):23-32.
  18. D'Urbano LE, Pellegrino K, Artesani MC, Donnanno S, Luciano R, Riccardi C, et al. Performance of a component-based allergen-microarray in the diagnosis of cow's milk and hen's egg allergy. Clin Exp Allergy. 2010;40(10):1561-70.
  19. Liu AH, Jaramillo R, Sicherer SH, Wood RA, Bock SA, Burks AW, et al. National prevalence and risk factors for food allergy and relationship to asthma: results from the National Health and Nutrition Examination Survey 2005-2006. J Allergy Clin Immunol. 2010;126(4):798-806 e13.
  20. Tedner G, Cilla S, Jon K, Eline BK, Magnus B, Kal-Hakon C, et al. Extract and molecular-based early infant sensitisation and associated factors – a PreventADALL study. Allergy (ahead of print). 2021
  21. Schmitz R, Ellert U, Kalcklosch M, Dahm S, Thamm M. Patterns of sensitization to inhalant and food allergens - findings from the German Health Interview and Examination Survey for Children and Adolescents. Int Arch Allergy Immunol. 2013;162(3):263-70.
  22. Yamamoto-Hanada K, Borres MP, Aberg MK, Yang L, Fukuie T, Narita M, et al. IgE responses to multiple allergen components among school-aged children in a general population birth cohort in Tokyo. World Allergy Organ J. 2020;13(2):100105.
  23. Burney P, Summers C, Chinn S, Hooper R, van Ree R, Lidholm J. Prevalence and distribution of sensitization to foods in the European Community Respiratory Health Survey: a EuroPrevall analysis. Allergy. 2010;65(9):1182-8.
  24. Venkataraman D, Erlewyn-Lajeunesse M, Kurukulaaratchy RJ, Potter S, Roberts G, Matthews S, et al. Prevalence and longitudinal trends of food allergy during childhood and adolescence: Results of the Isle of Wight Birth Cohort study. Clin Exp Allergy. 2018;48(4):394-402.
  25. AAAAI. What primary care givers need to know about the new guidlines for the diagnosis and management of food allergy in the US American Academy of Allergy, Asthma and Immunology. 2012.
  26. Ohtani K, Sato S, Syukuya A, Asaumi T, Ogura K, Koike Y, et al. Natural history of immediate-type hen's egg allergy in Japanese children. Allergol Int. 2016;65(2):153-7.
  27. Savage JH, Matsui EC, Skripak JM, Wood RA. The natural history of egg allergy. J Allergy Clin Immunol. 2007;120(6):1413-7.
  28. Caubet JC, Bencharitiwong R, Moshier E, Godbold JH, Sampson HA, Nowak-Wegrzyn A. Significance of ovomucoid- and ovalbumin-specific IgE/IgG(4) ratios in egg allergy. J Allergy Clin Immunol. 2012;129(3):739-47.
  29. Dang TD, Mills CE, Allen KJ. Determination of the clinical egg allergy phenotypes using component-resolved diagnostics. Pediatr Allergy Immunol. 2014;25(7):639-43.
  30. Savage J, Johns CB. Food allergy: epidemiology and natural history. Immunol Allergy Clin North Am. 2015;35(1):45-59.
  31. Koplin JJ, Dharmage SC, Ponsonby AL, Tang ML, Lowe AJ, Gurrin LC, et al. Environmental and demographic risk factors for egg allergy in a population-based study of infants. Allergy. 2012;67(11):1415-22.
  32. Dang TD, Peters RL, Koplin JJ, Dharmage SC, Gurrin LC, Ponsonby AL, et al. Egg allergen specific IgE diversity predicts resolution of egg allergy in the population cohort HealthNuts. Allergy. 2019;74(2):318-26.
  33. Sampson HA, Aceves S, Bock SA, James J, Jones S, Lang D, et al. Food allergy: a practice parameter update—2014. Journal of Allergy and Clinical Immunology. 2014;134(5):1016-25. e43.
  34. Wood RA. Allergic reactions to vaccines. Pediatr Allergy Immunol. 2013;24(6):521-6.
  35. Turner PJ, Southern J, Andrews NJ, Miller E, Erlewyn-Lajeunesse M, Investigators S-S. Safety of live attenuated influenza vaccine in young people with egg allergy: multicentre prospective cohort study. BMJ. 2015;351:h6291.
  36. NACI NACoI. Egg allergy and MMR vaccine: New recommendations from the National Advisory Committee on Immunization. Can J Infect Dis. 1996;7(5):289-90.
  37. Tariq SM, Matthews SM, Hakim EA, Arshad SH. Egg allergy in infancy predicts respiratory allergic disease by 4 years of age. Pediatric Allergy and Immunology. 2000;11(3):162-7.
  38. Ma X., Liang R., Xing Q., D. L-O. Can food processing produce hypoallergenic egg? J Food Sci. 2020;85(9):2635-44.
  39. Echeverria L, Martin-Munoz MF, Martorell C, Belver MT, Alonso Lebrero E, Zapatero L, et al. Clinical and immunological profile of children aged 5-9 years with persistent egg allergy before oral immunotherapy with egg. A multicenter, randomized controlled trial of the Spanish Society of Pediatric Allergy, Asthma and Clinical Immunology (SEICAP). Allergol Immunopathol (Madr). 2018;46(5):415-20.
  40. Komata T, Söderström L, Borres MP, Tachimoto H, Ebisawa M. The predictive relationship of food-specific serum IgE concentrations to challenge outcomes for egg and milk varies by patient age. Journal of allergy and clinical immunology. 2007;119(5):1272.
  41. Palosuo K, Kukkonen AK, Pelkonen AS, Mäkelä MJ. Gal d 1‐specific IgE predicts allergy to heated egg in Finnish children. Pediatric Allergy and Immunology. 2018;29(6):637-43.
  42. Ando H, Moverare R, Kondo Y, Tsuge I, Tanaka A, Borres MP, et al. Utility of ovomucoid-specific IgE concentrations in predicting symptomatic egg allergy. J Allergy Clin Immunol. 2008;122(3):583-8.
  43. Jessadapakorn W, Sangsupawanich P, Wootipoom N, Suddeaugrai O, Yuenyongviwat A. Component-resolved diagnostics in Thai children with cow's milk and egg allergy. Asian Pac J Allergy Immunol. 2017;35(4):179-85.
  44. Petrosino MI, Scaparrotta A, Marcovecchio ML, Panichi D, Rapino D, Attanasi M, et al. Usefulness of molecular diagnosis in egg allergic children. Arch Med Sci. 2018;14(1):132-7.
  45. Boyano Martinez T, Garcia‐Ara C, Diaz‐Pena J, Muñoz FM, Garcia Sanchez G, Esteban MM. Validity of specific IgE antibodies in children with egg allergy. Clinical & Experimental Allergy. 2001;31(9):1464-9.
  46. Roy-Ghanta S, Larosa DF, Katzka DA. Atopic characteristics of adult patients with eosinophilic esophagitis. Clin Gastroenterol Hepatol. 2008;6(5):531-5.
  47. Sampson HA. Utility of food-specific IgE concentrations in predicting symptomatic food allergy. Journal of Allergy and Clinical Immunology. 2001;107(5):891-6.
  48. Konstantinou GN, Giavi S, Kalobatsou A, Vassilopoulou E, Douladiris N, Saxoni-Papageorgiou P, et al. Consumption of heat-treated egg by children allergic or sensitized to egg can affect the natural course of egg allergy: hypothesis-generating observations. J Allergy Clin Immunol. 2008;122(2):414-5.
  49. Koplin JJ, Osborne NJ, Wake M, Martin PE, Gurrin LC, Robinson MN, et al. Can early introduction of egg prevent egg allergy in infants? A population-based study. J Allergy Clin Immunol. 2010;126(4):807-13.
  50. Caminiti L, Pajno GB, Crisafulli G, Chiera F, Collura M, Panasci G, et al. Oral Immunotherapy for Egg Allergy: A Double-Blind Placebo-Controlled Study, with Postdesensitization Follow-Up. J Allergy Clin Immunol Pract. 2015;3(4):532-9.
  51. Borres MP, Ebisawa M, Eigenmann PA. Use of allergen components begins a new era in pediatric allergology. Pediatric Allergy and immunology. 2011;22(5):454-61.
  52. WHO-IUIS. IUIS Allergen Nomenclature 2019 [27-November-2020]. Available from: http://www.allergen.org/search.php?Species=Gallus%20domesticus%20(G.%20gallus).
  53. Eigenmann AP, Caubet J-C, Muraro A. B11. ALLERGY TO EGG. In: Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, et al., editors. EAACI Molecular Allergology User's Guide2016.
  54. Palosuo K. KAK, Pelkonen A. S., and Makela M. J. Gal d 1-specific IgE predicts allergy to heated egg in Finnish children. Pediatr Allergy Immunol. 2018;29(6):637-43.
  55. Liu Y, Qiu N, Ma M. Comparative proteomic analysis of egg white proteins during the rapid embryonic growth period by combinatorial peptide ligand libraries. Poultry science. 2015;94(10):2495-505.
  56. Sato S, Yanagida N, Ebisawa M. How to diagnose food allergy. Current opinion in allergy and clinical immunology. 2018;18(3):214-21.
  57. Lunhui H, Yanhong S, Shaoshen L, Huijing B, Yunde L, Huiqiang L. Component resolved diagnosis of egg yolk is an indispensable part of egg allergy. Allergol Immunopathol (Madr). 2021;49(2):6-14.
  58. Haneda Y, Kando N, Yasui M, Kobayashi T, Maeda T, Hino A, et al. Ovomucoids IgE is a better marker than egg white-specific IgE to diagnose boiled egg allergy. J Allergy Clin Immunol. 2012;129(6):1681-2.
  59. Kattan JD, Wang J. Allergen component testing for food allergy: ready for prime time? Curr Allergy Asthma Rep. 2013;13(1):58-63.
  60. Ou K, Seow TK, Liang RC, Lee BW, Goh DL, Chua KY, et al. Identification of a serine protease inhibitor homologue in Bird's Nest by an integrated proteomics approach. Electrophoresis. 2001;22(16):3589-95.
  61. Mandallaz MM, de Weck AL, Dahinden CA. Bird-egg syndrome. Cross-reactivity between bird antigens and egg-yolk livetins in IgE-mediated hypersensitivity. Int Arch Allergy Appl Immunol. 1988;87(2):143-50.
  62. Walsh B, Elliott C, Baker R, Barnett D, Burley R, Hill D, et al. Allergenic cross-reactivity of egg-white and egg-yolk proteins. International Archives of Allergy and Immunology. 1987;84(3):228-32.