Allergenic molecules
To date, two allergenic molecules have been identified, characterized and published officially by the World Health Organization (WHO) and International Union of Immunological Societies’ (IUIS) Allergen Nomenclature Subcommittee for Brazil nut (14, 26). The table below provides detailed information on each of the allergenic protein identified by WHO/IUIS as of 25 January 2021:
Allergen |
Isoallergen |
Biochemical Name |
Molecular Weight (kDa) |
Allergenicity |
---|
Ber e 1 |
Ber e 1.0101 |
2S albumin |
9 |
- Major allergen (11, 22).
- Resistance to heat and pepsin digestion (6).
- 100% of Brazil nut-allergic patients with anaphylaxis showed binding on immunoblots (n=11) (27).
- Lesser immunoreactivity compared to Ber e 2 (11).
|
Ber e 2 |
Ber e 2.0101 |
11S globulin-like protein |
29 |
- Major allergen (11).
- Also referred to as excelsin (12).
- Specific IgE-binding was observed in 44% of Brazil nut-sensitized patients via Western blotting (12 out of 27) (28).
|
Allergen |
Isoallergen |
Biochemical Name |
Molecular Weight (kDa) |
Allergenicity |
---|
kDa: kilodaltons, IgE: Immunoglobulin E
According to the literature, it has been revealed that the phospholipids in Brazil nut oil could play an important role in the inherent property of Ber e 1 to elicit allergic reactions (29).
Biomarkers of severity
The allergen Ber e 1 has been found to be thermostable and hence, roasting at 100°C for 20 min was not able to alleviate the allergenicity of the molecule. Ber e 1 has also been found to resist pepsin digestion. Furthermore, it has been reported that even alkaline treatment of the allergen was shown to be ineffective in reducing the immunoreactivity (6).
Ber e 1-sIgE may serve as a good diagnostic marker in identifying true Brazil nut allergy as compared to sIgE to whole Brazil nut extract (12, 22).
Cross-reactivity
Several studies have demonstrated that individuals reactive to one tree nut may be sensitized to the other, and this may be due to the cross-reactivity among their pan-allergens or nut storage proteins (30).
A survey was conducted to determine the true cross-reactivity between different tree-nut antigens. The tree nuts were categorized into two groups of cross-reactive nuts – strong: walnut, pecan, and hazelnut and moderate: Brazil nut, hazelnut, cashew, pistachio, and almond. The immunological cross-reactivity has been reported between Brazil nut, hazelnut, cashew, pistachio, and almond (moderate cross-reactive group), which may be because they belong to the same subclass i.e. Rosidae. However, cross-reactivity has also been observed among the members of the two groups of cross-reactive nuts i.e. between pecan and Brazil nut and between walnut and Brazil nut (31).
Further, the Pronuts study in Europe found the percentage of co-existent nut allergies with Brazil nut to be 70% with walnut (7 out of 10), 40% each with hazelnut, cashew, macadamia, pecan (4 out of 10), 30% with sesame (3 out of 10), 20% each with pistachio and peanuts (2 out of 10) and 10% with pine nut (1 out of 10) among 10 Brazil-nut allergic patients (19).
Additionally, IgE-mediated cross-reactivity between Brazil nut and walnut and pecan has also been found (30). In a study, cross-reactivity patterns were identified by xMAP multiplex food allergen detection assay, that found Brazil nut exhibiting cross-reactivity with cashew, hazelnut, and walnut (10).
It has been speculated that peanut-allergic individuals may be allergic to Brazil nut too (4).
Furthermore, moderate degree of co-sensitization exists between Brazil nut and coconut. This was supported by a retrospective analysis study that showed 58% of co-sensitization between Brazil nut and coconut, which was found to be statistically significant (p<0.001) (32).
Structural identity and similarity have been proposed between the 2S albumins of several foods. It has been reported that the 2S albumin of cocoa seed was found to share 44% of identity and 69% of similarity with the 2S albumin of Brazil nut (33, 34). Furthermore, cross-reactivity as a result of structural homology may be present between the 2S albumin of Brazil nut (Ber e 1) and other plant species, such as cottonseed, sunflower, rapeseed, castor bean, and sesame (35).