Whole Allergen
f245 Egg
f245 Egg Scientific Information
Type:
Whole Allergen
Display Name:
Egg
Route of Exposure:
Ingestion
Family:
Phasianidae
Species:
Gallus gallus
Latin Name:
Gallus domesticus
Summary
Hen’s egg (Gallus gallus domesticus) is a protein-rich food source and is usually consumed as raw, cooked, or processed. Both egg white and egg yolk are rich in proteins, but egg white is regarded as more allergenic. The egg has been considered one of the most common food allergy sources, particularly in infants and children. Around 0.5 – 2.5 % of young children within a normal population have been found to be allergic to eggs. However, the prevalence of egg allergy in adults is less than 0.25 %. Egg sensitization, having IgE antibodies to egg, is a prerequisite for egg allergy. Egg sensitization is more prevalent than egg allergy, and the prevalence rate has been reported to be up to 6 % depending on age. Most egg-allergic children have the ability to develop tolerance to egg proteins over time. A patient's highest recorded egg sIgE is significantly related to the persistence of egg allergy. A higher egg sIgE value denotes a more extended period to develop tolerance. Clinical manifestations of egg allergy include mild to severe reactions like skin reactions (erythema, urticaria, angioedema), gastrointestinal symptoms (vomiting, abdominal pain), respiratory conditions (rhinitis, conjunctivitis, cough, bronchospasm, breathing difficulty), and anaphylaxis. Therefore, egg-allergic individuals are usually recommended to avoid egg and egg-based food products strictly. Moreover, egg allergy children are often falsely denied influenza or MMR vaccination as the vaccine might contain a small amount of egg protein. However, several studies and guidelines have claimed these vaccines to be safe for children with egg allergies.
An oral food challenge is usually considered the gold standard for diagnosing egg allergy. Egg sIgE testing can also aid in diagnosing egg allergy and is predictive of allergy outcome; therefore, it should be utilized in counseling patients on prognosis. Furthermore, egg allergy or sensitization has been found to elevate the risk of developing sensitization to aeroallergens (such as grass pollens, house dust mites, cats, and dogs).
Early introduction of egg into the infants’ diet has been found to be beneficial in developing tolerance to the egg; it is presently studied as a means of primary prevention of egg allergy. Additionally, oral immunotherapy (for egg) is also performed in some countries on egg-allergic patients who fail to develop natural tolerance to the egg.
Allergen
Nature
Egg (Gallus gallus) is a food product with high nutrition content, suitable and popular among individuals of all ages (children, adults, elderly). A combination of perfect nutritional balance, easy digestibility, economic and widespread accessibility makes egg a vital part of the human diet (1).
Hen’s egg is regarded as one of the essential poultry products and is a significant protein source for consumers globally (2). The egg is reported to contain both macro and micronutrients required for embryonic development until hatching the chicks (from the egg) (1).
Egg white is mainly composed of water and lacks lipids, whereas egg yolk is a rich source of lipids, vitamins, and minerals (1).
Protein from egg white is potentially more allergenic compared to protein from egg yolk (3). Egg white possesses 23 different proteins, making it a potential source of allergen. The significant allergens found in egg white include ovomucoid (Gal d 1), ovalbumin (Gal d 2), ovotransferrin (Gal d 3), and lysozyme (Gal d 4) (4). While alpha-livetin (Gal d 5) and YGP42 (Gal d 6) have been identified as allergens in egg yolk (5).
Globally, eggs have been extensively utilized in food products like bread, pasta, and others (3). 
Habitat
Hen’s eggs are regarded as fresh up to 28 days post laying. Room temperature or preferably refrigeration is considered ideal for the storage of shelled eggs. Variation in storage condition and duration can induce physiochemical alterations and further protein denaturation in eggs (1). 
Taxonomy
An in-vitro study confirmed high cross-reactivity between hen’s egg (Gallus domesticus) with egg white from turkey, duck, goose, and seagull (6). 
Taxonomic tree of Hen’s Egg (2)
Domain Eukaryota
Kingdom Animalia
Phylum Chrodata
Class Aves
Order Galliformes
Family Phasianidae
Genus Gallus
Species Gallus gallus
Taxonomic tree of Hen’s Egg (2)
Tissue
The egg comprises egg white and egg yolk, which are protein-rich edible parts. The average protein level of a whole raw fresh egg is about 12.5 g/100 g (1). Well-cooked eggs are considered safer for consumption than raw or undercooked eggs (7).
During the cooking process, egg proteins are claimed to undergo significant conformational modifications, leading to protein denaturation. This process of protein denaturation may lead to the inactivation of antinutritional factors (1). 
Epidemiology
Worldwide distribution
The egg has been reported as one of the common inducers of food allergy prevalent in industrialized countries (8). According to a study, around 0.5-2.5% of young children have been observed to be affected with egg allergy (9, 10). A Norway-based birth cohort study reported the prevalence of egg allergy among children (2½ years) to be 1.6% (11). In another recent study (EuroPrevall), self-reported hen’s egg allergy was observed in 9.9% of 16,935 children (7-10 years) (12). According to a population-based study in the US, the overall prevalence of egg allergy among children (0-17 years) was found to be 0.8% (304/38,480) (13). Moreover, an Australian population study identified the prevalence of raw egg allergy among 12-month old infants to be 9.0% (14). Additionally, in Japan, hen’s egg has been reported to be the most frequent cause of food allergy (15). Furthermore, a US-based cohort study also detected the prevalence of egg allergy to be approximately <0.25% (from graphical representation) in 4425 adults (20-60+ years) (16).
Sensitization to the egg means that IgE antibodies are produced towards the egg proteins and are a prerequisite for an IgE-mediated egg allergy. Egg sensitization is more prevalent than egg allergy, and IgE antibodies have been detected already in very young age groups. Tedner et al found in a Nordic population-based birth-cohort study the prevalence of egg sensitization among 3-month old infants to be 3.7% (41/1102) (17). Another cohort study in Sweden on 2336 children (4 years) claimed the prevalence of egg sensitization as 5% (112/2336) (18). Moreover, a German-based cohort study confirmed the highest sensitization towards hen’s egg (around 6%) in infants at 1 year of age (19). Another German-based population study conducted on 14,836 children and adolescents (3-17 years) found the overall sensitization rate towards egg white was 5% (20). Additionally, a European cohort study found an overall prevalence of egg sensitization to be 0.4% among the adult (20-39 years) population (21). Furthermore, a US-based cohort study reported the prevalence of egg sensitization in the adult population to be ranging between 2.1%  to 3.9% among 4425 adults (20-60+ years) (16). 
Natural History of Egg Allergy
Food allergy during childhood could be a transitory condition, although in some cases, it is found to be a persistent one. Hen’s egg has been reported to be one of the frequent food allergens affecting children during the first ten years of life (22). As per the American Academy of Allergy, Asthma & Immunology (AAAAI) guidelines, egg allergy has been claimed to get resolved naturally with aging (23).  Moreover, the egg sIgE level has been regarded as one of the crucial predictive factors for the resolution of egg allergy (10, Savage 2007).
A study conducted on 881 egg-allergic patients (10-23 months old) reported the development of tolerance towards eggs by the age of 4 years in 4% cases and by 16 years in 68% cases. A gradual decline in egg sIgE level was evident with aging among patients with persistent egg-allergy; in contrast, a sharp decline in egg sIgE was found in egg-allergy resolved (tolerant) patients from this study  (24). Similarly, another study conducted on 226 egg-allergic children (<6 years) reported tolerance development in 30% (66/226) of children by the age of 3 years, while in 73% (164/226) of children by 6 years (15). Furthermore, as per another study, the probability of developing egg tolerance (based on the level of sIgE) in children (<4 years) was found to be 0.52 with a 50% reduction and 0.95 with a 99% reduction in sIgE level (for 12 months) (25).
Risk factors
Egg allergy is associated with atopic dermatitis, asthma, rhinitis, and other food allergies (such as peanut) (10). Clinical allergy towards eggs has been found to be associated with risk factors like male gender, young age, ethnicity/race (26), and family history (27).
Children with egg allergy are often denied influenza vaccination because the vaccine may contain a small amount of egg protein. However, recent studies have demonstrated that children with even severe egg allergy can safely receive the influenza vaccine. Therefore, based on this outcome, the current guidelines recommend routine immunization (influenza vaccination) of such egg-allergic patients without undergoing any testing or special considerations  (28). Moreover, as per the recommendations of vaccination-specific guidelines, the influenza vaccine can be received under usual clinical settings (29). A study conducted on egg-allergic patients immunized with live attenuated influenza vaccine reported no systemic reactions (allergic) or anaphylaxis among these patients post-immunization (30).
National Advisory Committee on Immunization (NACI) has also Sdeemed patients with egg allergy suitable for receiving the Measles Mumps Rubella (MMR) vaccination. According to this guideline, the immunization may be administered without prior testing but under proper guidance and adequate facility (31). 
Pediatric issues
A significant correlation exists between sensitization towards outdoor/indoor aeroallergens (like grass pollens, house dust mites, cat, dog) and egg allergy during infancy (32, 33).
Additionally, the coexistence of egg allergy and eczema at infancy has also been reported as a predictive indicator of aeroallergen sensitization and respiratory allergies among children (4 years) (33). 
Environmental Characteristics
Worldwide distribution
Hen’s egg is consumed globally in various forms like raw, semi-cooked (poached or soft-boiled eggs) (1), completely cooked (hard-boiled, scrambled, baked, or fried) (5) or as an additive in products like bread, waffles, cakes, and others (3). Additionally, eggs are also utilized in the cosmetic, pharmaceutical (especially vaccine preparation), and beverage refining (wine and beer) industry (5). 
Route of Exposure
Main
The route of exposure is through the ingestion (consumption) of eggs or products containing eggs (34). 
Clinical Relevance
Egg-allergic patients might experience a range of symptoms or reactions after consumption of egg or egg-based products. Based on the severity, these reactions can be further categorized as mild reactions (vomiting, cough, conjunctivitis, rhinitis, localized urticaria, erythema), moderate reactions (mild bronchospasm, facial angioedema, generalized urticaria), and severe reactions (anaphylactic shock, hypotension, breathing difficulty, severe bronchospasm) (34). 
Anaphylaxis
Egg-allergic patients with asthma are at high risk of manifesting anaphylaxis on egg exposure (7). An oral food challenge (OFC) study on 374 egg white-allergic patients reported anaphylaxis among 2% (8/374) of the patients (35). Another study in Korea on 363 infants (0 – 2 years old) with anaphylaxis found hen’s egg as a common food allergen, affecting 21.9% (74/338) of the infants (36). Additionally, anaphylaxis was claimed to be higher among children with prolonged egg allergy (15).  
Gastrointestinal (GI) symptoms
GI symptoms are mostly seen as immediate hypersensitivity reactions in egg-allergic individuals (7). According to a study, 21% (78/881) of the patients showed GI reactions (like abdominal pain, bloody stool, diarrhea, vomiting) as initial symptoms of egg allergy (24). As per another OFC study, 9% (34 out of 374) of egg white-allergic children exhibited GI reactions (35).
Skin reactions and Atopic dermatitis (AD)
Skin reactions like urticaria or angioedema are reported as IgE-mediated common allergic reactions prevalent in children (within minutes to a few hours) post-consumption of eggs (7). A study conducted on 881 egg-allergic patients exhibited skin reactions (like a non-eczematous rash, angioedema, and urticaria) as initial egg allergy symptoms in 58% (219/881) of the patients. Additionally, eczema was also observed in 18% (69/881) of the egg-allergic patients (24). Another study (OFC with egg white) found cutaneous reactions in 98% (368/374) of patients (35).
A higher prevalence of AD has been found among egg sensitized children compared to children not sensitized (32). Moreover,  an OFC study reported a positive clinical reaction towards hen’s egg in 62% (19/64) of children with AD (37). 
Asthma
A study reported egg as one of the typical food allergens prevalent in 185 of the 365 asthmatic children with food allergy (38).
Lower respiratory symptoms and mucous membrane reactions were observed in 6% (21 out of 374) and 2% (7 out of 374) of egg white-sensitized children in an OFC study (35).
Diagnostics Sensitization
Challenge tests
OFC is considered the gold standard for diagnosing egg allergy. However, this mode of diagnosis is often regarded as time extensive, resource-consuming, and potentially risky (39, 40).
In vitro diagnostics
Egg sIgE testing is used as an aid in diagnosing egg allergy and to monitor the disease development. The sIgE diagnostic values have been reported to be beneficial for physicians in deciding the necessity of an egg challenge while estimating its potential risk on the patients’ health. Furthermore, an age-specific correlation has been observed between egg sIgE levels and OFC outcome; younger children usually exhibit reactivity to even at low egg sIgE levels in contrast to the older children (35).
Periodic monitoring of the absolute values of egg sIgE antibody level combined with the clinical history of egg allergy can accurately diagnose egg allergy (41). 
Prevention and Therapy
Allergen immunotherapy
A study was conducted on 30 egg-allergic children (≥4 years) to estimate the efficacy of oral immunotherapy (OIT)-egg desensitization and evaluate the maintenance of tolerance. All the patients (n=16) attained desensitization towards egg after 4 months of therapy, while 31% were reported to retain tolerance towards egg even after 3 months of avoiding egg consumption (44).
An exploratory study was conducted to identify the reason for maintaining egg intake in patients post achieving desensitization through OIT. The study involved three groups, which included the non-egg-allergic group, pre-/post-egg OIT group, and naturally egg-allergy outgrown group. Higher sIgE levels were evident in post-egg OIT participants compared to the naturally egg-allergy outgrown group. Therefore, the study recommended egg intake in patients post-OIT to maintain desensitization (45).
Prevention strategies
Complete avoidance of egg and egg-based products from the diet is considered the possible treatment option for egg allergy (34); however, this could be challenging as chances of accidental intake of egg or egg-based products are usually quite high (10).
Patients allergic to heat-labile egg protein (like ovalbumin) might be tolerant towards cooked, boiled, or fried form of eggs (34). Therefore, differential avoidance is typically recommended for egg-allergic patients based on their reactivity towards raw or cooked eggs.  Moreover, reactivity towards raw or slightly cooked eggs has been found to be more prevalent compared to extensively processed egg-based food (42).
Early introduction of eggs into the diet has been found to potentially beneficial in the prevention of egg allergy (8). In a study conducted on 231 infants allergic to egg, early introduction of egg (4-6 months age) was found to be correlated with a reduced risk of egg allergy. In contrast, late introduction of an egg-based diet (at 10 months) might increase the risk of developing egg allergy (43).
Molecular Aspects
Allergenic molecules
Egg white possesses 23 distinct proteins and is considered a vital allergen source from egg (4). However, only 6 allergenic egg proteins (listed below in the table) from G. domesticus (Hen’s egg) have been officially recognized in the IUIS database (46). 
Allergens Molecular Weight (kDa) Biochemical name Allergenicity
Gal d 1 28 Ovomucoid
  • Gal d 1 is a strongly allergenic, heat-resistant protein (47) [p169] and contributes 11% of protein content in egg white (48).
  • According to an OFC study, 43.5% (20/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 1 (40).
Gal d 2 44 Ovalbumin
  • Gal d 2 is heat-unstable and therefore bears the risk of manifesting clinical reactivity towards the raw or slightly heated egg (47) [p169]; it contributes 54% of protein content in egg white (48).
  • According to an OFC study, 52.1% (24/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 2 (40).
Gal d 3 78 Ovotransferrin
  • Gal d 3 is thermo-labile and can manifest the risk of clinical reactivity towards raw or slightly heated egg (47)[p169]; it contributes 12% of the protein content in egg white (48).
  • According to an OFC study, 13.0% (6/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 3 (40).
Gal d 4 14 Lysozyme C
  • Gal d 4 contributes 3% of protein content in egg white (48).
  • According to an OFC study, 36.9% (17/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 4 (40).
Gal d 5 69 Serum albumin
  • Gal d 5 (serum albumin) is highly prevalent in egg yolk, and a similar type of serum albumin is also found in bird tissues (like muscle tissue).
  • Individuals sensitized towards serum albumin in hen’s egg may also exhibit poultry meat allergy due to cross-reactivity (49, 50).
  • According to an OFC study, 4.3% (2/46) of the patients (0.7-15.1 years) exhibited sIgE reactivity towards Gal d 5 (40)
Gal d 7 35 YGP42
  • Gal d 6 is a thermo-stable, pepsin-digestible protein found in egg yolk.
  • 18% (5/27) of sera from 27 EA patients (2-74 years) was found to show sIgE binding towards Gal d 6 (51).
Allergens Molecular Weight (kDa) Biochemical name Allergenicity
sIgE: specific Immunoglobulin E, kDa: kilodaltons, EA: Egg Allergy, ES: Egg-sensitized
Apart from the allergens mentioned above, Ovomucin (165 kDa; heavily glycosylated protein), an allergenic protein containing a trypsin inhibitor-like domain with potent antiviral activity, is also found in egg-white (47).
Further, other additional allergenic components found in egg yolk include phosvitin (transferase protein, 35 kDa), Apovitellenins I (very low-density lipoprotein, 9.5 kDa), and Apovitellenins VI (orapoprotein B, 170 kDa) (47).
Biomarkers of severity
As per a study, the best diagnostic biomarkers for raw and cooked egg allergy are distinct. Studies have found ovomucoid (Gal d 1) sIgE as a superior biomarker for cooked egg allergy (52, 53). In contrast, ovalbumin (Gal d 2) sIgE has been claimed as the best marker for diagnosing both raw and cooked egg allergy (53). 
Cross-reactivity
Hen’s egg exhibited high cross-reactivity with egg white from turkey, duck, goose, and seagull (6). Moreover, the presence of various avian serum albumins could be responsible for cross-reactivity between bird feathers and egg yolk, according to a study (50). Furthermore, livetin (allergenic protein) mediated cross-reactivity between hen’s egg proteins and bird dander has also been reported (54). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Magnus Borres
Last reviewed: March 2021
References
  1. Rehault-Godbert S, Guyot N, Nys Y. The Golden Egg: Nutritional Value, Bioactivities, and Emerging Benefits for Human Health. Nutrients. 2019;11(3).
  2. Al-Nasser A, Al-Khalaifa H, Al-Saffar A, Khalil F, Al-Bahouh M, Raghbeb G, et al. Overview of chicken taxonomy and domestication. World's Poultry Science Journal. 2007;63.
  3. Verhoeckx KCM, Vissers YM, Baumert JL, Faludi R, Feys M, Flanagan S, et al. Food processing and allergenicity. Food Chem Toxicol. 2015;80:223-40.
  4. Borres MP, Maruyama N, Sato S, Ebisawa M. Recent advances in component resolved diagnosis in food allergy. Allergol Int. 2016;65(4):378-87.
  5. Ma X., Liang R., Xing Q., D. L-O. Can food processing produce hypoallergenic egg? J Food Sci. 2020;85(9):2635-44.
  6. Langeland T. A clinical and immunological study of allergy to hen's egg white. VI. Occurrence of proteins cross-reacting with allergens in hen's egg white as studied in egg white from turkey, duck, goose, seagull, and in hen egg yolk, and hen and chicken sera and flesh. Allergy. 1983;38(6):399-412.
  7. Caubet JC, Wang J. Current understanding of egg allergy. Pediatr Clin North Am. 2011;58(2):427-43, xi.
  8. De Martinis M, Sirufo MM, Suppa M, Ginaldi L. New perspectives in food allergy. International journal of molecular sciences. 2020;21(4):1474.
  9. Rona RJ, Keil T, Summers C, Gislason D, Zuidmeer L, Sodergren E, et al. The prevalence of food allergy: a meta-analysis. J Allergy Clin Immunol. 2007;120(3):638-46.
  10. Sicherer SH, Wood RA, Vickery BP, Jones SM, Liu AH, Fleischer DM, et al. The natural history of egg allergy in an observational cohort. J Allergy Clin Immunol. 2014;133(2):492-9.
  11. Eggesbø M, Botten G, Halvorsen R, Magnus P. The prevalence of allergy to egg: a population‐based study in young children. Allergy. 2001;56(5):403-11.
  12. Lyons SA, Clausen M, Knulst AC, Ballmer-Weber BK, Fernandez-Rivas M, Barreales L, et al. Prevalence of Food Sensitization and Food Allergy in Children Across Europe. J Allergy Clin Immunol Pract. 2020;8(8):2736-46 e9.
  13. Gupta RS, Springston EE, Warrier MR, Smith B, Kumar R, Pongracic J, et al. The prevalence, severity, and distribution of childhood food allergy in the United States. Pediatrics. 2011;128(1):e9-17.
  14. Osborne NJ, Koplin JJ, Martin PE, Gurrin LC, Lowe AJ, Matheson MC, et al. Prevalence of challenge-proven IgE-mediated food allergy using population-based sampling and predetermined challenge criteria in infants. J Allergy Clin Immunol. 2011;127(3):668-76 e1-2.
  15. Ohtani K, Sato S, Syukuya A, Asaumi T, Ogura K, Koike Y, et al. Natural history of immediate-type hen's egg allergy in Japanese children. Allergol Int. 2016;65(2):153-7.
  16. Liu AH, Jaramillo R, Sicherer SH, Wood RA, Bock SA, Burks AW, et al. National prevalence and risk factors for food allergy and relationship to asthma: results from the National Health and Nutrition Examination Survey 2005-2006. J Allergy Clin Immunol. 2010;126(4):798-806 e13.
  17. Tedner G, Cilla S, Jon K, Eline BK, Magnus B, Kal-Hakon C, et al. Extract and molecular-based early infant sensitisation and associated factors – a PreventADALL study. Allergy (ahead of print). 2021
  18. Ostblom E, Lilja G, Ahlstedt S, van Hage M, Wickman M. Patterns of quantitative food-specific IgE-antibodies and reported food hypersensitivity in 4-year-old children. Allergy. 2008;63(4):418-24.
  19. Kulig M, Bergmann R, Klettke U, Wahn V, Tacke U, Wahn U. Natural course of sensitization to food and inhalant allergens during the first 6 years of life. J Allergy Clin Immunol. 1999;103(6):1173-9.
  20. Schmitz R, Ellert U, Kalcklosch M, Dahm S, Thamm M. Patterns of sensitization to inhalant and food allergens - findings from the German Health Interview and Examination Survey for Children and Adolescents. Int Arch Allergy Immunol. 2013;162(3):263-70.
  21. Burney P, Summers C, Chinn S, Hooper R, van Ree R, Lidholm J. Prevalence and distribution of sensitization to foods in the European Community Respiratory Health Survey: a EuroPrevall analysis. Allergy. 2010;65(9):1182-8.
  22. Venkataraman D, Erlewyn-Lajeunesse M, Kurukulaaratchy RJ, Potter S, Roberts G, Matthews S, et al. Prevalence and longitudinal trends of food allergy during childhood and adolescence: Results of the Isle of Wight Birth Cohort study. Clin Exp Allergy. 2018;48(4):394-402.
  23. AAAAI. What primary care givers need to know about the new guidlines for the diagnosis and management of food allergy in the US American Academy of Allergy, Asthma and Immunology. 2012.
  24. Savage JH, Matsui EC, Skripak JM, Wood RA. The natural history of egg allergy. Journal of Allergy and Clinical Immunology. 2007;120(6):1413-7.
  25. Shek LP, Soderstrom L, Ahlstedt S, Beyer K, Sampson HA. Determination of food specific IgE levels over time can predict the development of tolerance in cow's milk and hen's egg allergy. J Allergy Clin Immunol. 2004;114(2):387-91.
  26. Liu AH, Jaramillo R, Sicherer SH, Wood RA, Bock SA, Burks AW, et al. National prevalence and risk factors for food allergy and relationship to asthma: results from the National Health and Nutrition Examination Survey 2005-2006. J Allergy Clin Immunol. 2010;126(4):798-806.e13.
  27. Koplin JJ, Dharmage S, Ponsonby AL, Tang M, Lowe A, Gurrin LC, et al. Environmental and demographic risk factors for egg allergy in a population‐based study of infants. Allergy. 2012;67(11):1415-22.
  28. Sampson HA, Aceves S, Bock SA, James J, Jones S, Lang D, et al. Food allergy: a practice parameter update—2014. Journal of Allergy and Clinical Immunology. 2014;134(5):1016-25. e43.
  29. Wood RA. Allergic reactions to vaccines. Pediatr Allergy Immunol. 2013;24(6):521-6.
  30. Turner PJ, Southern J, Andrews NJ, Miller E, Erlewyn-Lajeunesse M. Safety of live attenuated influenza vaccine in young people with egg allergy: multicentre prospective cohort study. Bmj. 2015;351.
  31. NACI NACoI. Egg allergy and MMR vaccine: New recommendations from the National Advisory Committee on Immunization. Can J Infect Dis. 1996;7(5):289-90.
  32. Nickel R, Kulig M, Forster J, Bergmann R, Bauer CP, Lau S, et al. Sensitization to hen's egg at the age of twelve months is predictive for allergic sensitization to common indoor and outdoor allergens at the age of three years. J Allergy Clin Immunol. 1997;99(5):613-7.
  33. Tariq SM, Matthews SM, Hakim EA, Arshad SH. Egg allergy in infancy predicts respiratory allergic disease by 4 years of age. Pediatric Allergy and Immunology. 2000;11(3):162-7.
  34. Echeverria L, Martin-Munoz MF, Martorell C, Belver MT, Alonso Lebrero E, Zapatero L, et al. Clinical and immunological profile of children aged 5-9 years with persistent egg allergy before oral immunotherapy with egg. A multicenter, randomized controlled trial of the Spanish Society of Pediatric Allergy, Asthma and Clinical Immunology (SEICAP). Allergol Immunopathol (Madr). 2018;46(5):415-20.
  35. Komata T, Soderstrom L, Borres MP, Tachimoto H, Ebisawa M. The predictive relationship of food-specific serum IgE concentrations to challenge outcomes for egg and milk varies by patient age. J Allergy Clin Immunol. 2007;119(5):1272-4.
  36. Jeon YH, Lee S, Ahn K, Lee SY, Kim KW, Kim HH, et al. Infantile Anaphylaxis in Korea: a Multicenter Retrospective Case Study. J Korean Med Sci. 2019;34(13):e106.
  37. Breuer K, Heratizadeh A, Wulf A, Baumann U, Constien A, Tetau D, et al. Late eczematous reactions to food in children with atopic dermatitis. Clin Exp Allergy. 2004;34(5):817-24.
  38. Gaffin JM, Sheehan WJ, Morrill J, Cinar M, Borras Coughlin IM, Sawicki GS, et al. Tree nut allergy, egg allergy, and asthma in children. Clinical pediatrics. 2011;50(2):133-9.
  39. Petrosino MI, Scaparrotta A, Marcovecchio ML, Panichi D, Rapino D, Attanasi M, et al. Usefulness of molecular diagnosis in egg allergic children. Arch Med Sci. 2018;14(1):132-7.
  40. D'Urbano LE, Pellegrino K, Artesani MC, Donnanno S, Luciano R, Riccardi C, et al. Performance of a component-based allergen-microarray in the diagnosis of cow's milk and hen's egg allergy. Clin Exp Allergy. 2010;40(10):1561-70.
  41. Sampson HA. Utility of food-specific IgE concentrations in predicting symptomatic food allergy. Journal of Allergy and Clinical Immunology. 2001;107(5):891-6.
  42. Ando H, Moverare R, Kondo Y, Tsuge I, Tanaka A, Borres MP, et al. Utility of ovomucoid-specific IgE concentrations in predicting symptomatic egg allergy. J Allergy Clin Immunol. 2008;122(3):583-8.
  43. Koplin JJ, Osborne NJ, Wake M, Martin PE, Gurrin LC, Robinson MN, et al. Can early introduction of egg prevent egg allergy in infants? A population-based study. Journal of Allergy and Clinical Immunology. 2010;126(4):807-13.
  44. Caminiti L, Pajno GB, Crisafulli G, Chiera F, Collura M, Panasci G, et al. Oral Immunotherapy for Egg Allergy: A Double-Blind Placebo-Controlled Study, with Postdesensitization Follow-Up. J Allergy Clin Immunol Pract. 2015;3(4):532-9.
  45. Andorf S, Bunning B, Tupa D, Cao S, Long AJ, Borres MP, et al. Trends in egg specific immunoglobulin levels during natural tolerance and oral immunotherapy. Allergy. 2020;75(6):1454-6.
  46. WHO-IUIS. IUIS Allergen Nomenclature 2019 [27-November-2020]. Available from: http://www.allergen.org/search.php?allergenname=&allergensource=Gallus+domesticus+%28G.+gallus%29&TaxSource=&TaxOrder=&foodallerg=all&bioname=.
  47. Eigenmann AP, Caubet J-C, Muraro A. B11. ALLERGY TO EGG. In: Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, et al., editors. EAACI Molecular Allergology User's Guide2016.
  48. Palosuo K. KAK, Pelkonen A. S., and Makela M. J. Gal d 1-specific IgE predicts allergy to heated egg in Finnish children. Pediatr Allergy Immunol. 2018;29(6):637-43.
  49. Inomata N, Kawano K, Aihara M. Bird-egg syndrome induced by alpha-livetin sensitization in a budgerigar keeper: Successful induction of tolerance by avoiding exposure to avians. Allergol Int. 2019;68(2):282-4.
  50. Hemmer W, Klug C, Swoboda I. Update on the bird-egg syndrome and genuine poultry meat allergy. Allergo journal international. 2016;25(3):68-75.
  51. Amo A, Rodríguez-Pérez R, Blanco J, Villota J, Juste S, Moneo I, et al. Gal d 6 is the second allergen characterized from egg yolk. J Agric Food Chem. 2010;58(12):7453-7.
  52. Borres MP, Ebisawa M, Eigenmann PA. Use of allergen components begins a new era in pediatric allergology. Pediatric Allergy and immunology. 2011;22(5):454-61.
  53. Sato S, Yanagida N, Ebisawa M. How to diagnose food allergy. Current opinion in allergy and clinical immunology. 2018;18(3):214-21.
  54. Mandallaz MM, de Weck AL, Dahinden CA. Bird-egg syndrome. Cross-reactivity between bird antigens and egg-yolk livetins in IgE-mediated hypersensitivity. Int Arch Allergy Appl Immunol. 1988;87(2):143-50.