Whole Allergen
f313 Anchovy
f313 Anchovy Scientific Information
Type:
Whole Allergen
Display Name:
Anchovy
Family:
Engraulidae
Latin Name:
Engraulis encrasicolus
Other Names:
Anchovy
Summary
The names Anchovis or Ansjovis are used in Germany and Scandinavia for spice-cured sprat (Sprattus sprattus) or small herring.
Environmental Characteristics
Other topics
The anchovy is usually around 12-16 cm in length. They are found in the Atlantic, Mediterranean and Black Seas and migrate to the North Sea. The names Anchovis or Ansjovis are used in Germany and Scandinavia for spicecured sprat (Sprattus sprattus) or small herring.  See also sardine, f308.
Clinical Relevance
Anchovy has a high content of histamine. Only 35% of a group of codfish sensitive children showed sensitivity to anchovy (1). In another study, skin testing with a commercial anchovy extract was reported to give false positive results in 75% of non-atopic controls (2), presumably because of the high content of histamine (3).
Compiled By
Last Reviewed-  April 2022
References
  1. de Martino, M; Novembre, E; Galli, L; Marco, A; Botarelli, P; Marano, E; Vierucci, Allergy to different fish species in cod-allergic children: In vivo and in vitro studies. J Allergy Clin Immunol; 1990; 86: 909-914.
  2. Helbling, A; McCants, ML; Musmand, Schwartz, HJ; Lehrer, SB. Immunopathogenesis of fish allergy: identification of fish-allergic adults by skin test and radio-allergosorbent test. Ann Allergy, Asthma Immunol; 1996; 77: 48-54.
  3. Williams, PB; Nolte, H; Dolen, WK; et al. The histamine content of allergen extracts. J Allergy Clin Immunol; 1992; 89: 738-45.