Whole Allergen
g13 Velvet grass
g13 Velvet grass Scientific Information
Type:
Whole Allergen
Display Name:
Velvet grass
Route of Exposure:
Inhalation of pollen
Family:
Poaceae (Gramineae)
Species:
Holcus lanatus
Latin Name:
Holcus lanatus
Other Names:
Common velvet grass, creeping soft grass, meadow softgrass, Yorkshire fog
Summary
Holcus lanatus is a perennial, hairy, tufted grass that is common in temperate regions worldwide. High percentages of individuals (>77%) with seasonal allergic rhinitis have shown positive reactions to cutaneous provocation with H. lanatus pollen. Four allergenic molecules and epitopes have been characterized from H. lanatus pollen, including a Group 1 beta-expansin (Hol l 1), a Group 5 ribonuclease (Hol l 5), a Group 4 berberine bridge enzyme (Hol l 4), and a Group 2 protein with unknown biological function (Hol l 2). Hol l 1, Hol l 5, and to a lesser extent Hol l 4 have shown a high degree of cross-reactivity with homologs from other Pooideae subfamily grasses.
Allergen
Nature
Velvet grass is a perennial, hairy, tufted grass that grows 0.5 to 1.0 m tall (1, 2). Soft velvety leaves up to 20 cm long are the most consistent identifier for H. lanatus (2). H. lanatus has a purplish anthocyanin coloration on the panicles and veins of the sheaths (2). H. lanatus flowers from June to September in Europe and from May to October along the eastern coast of North America (1, 2). Velvet grass is wind-pollinated and produces abundant seed depending on plant age and the time at which flowers emerge (2). Common velvet grass (H. lanatus) has been reported to hybridize with creeping velvet grass (H. mollis) (1).
Taxonomy     
Taxonomic tree of Holcus lanatus (Velvet grass) (2)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Monocotyledonae
Family
Poaceae
Genus
Holcus.
Taxonomic tree of Holcus lanatus (Velvet grass) (2)
Environmental Characteristics
Living environment
Velvet grass has a wide climatic range and is commonly found on wet, swampy, poorly drained, or waterlogged soils within pastures, roadsides, waste ground or open woodland (2). Plants are relatively deep-rooted and can form dense networks of surface roots (2). In Brazil, H. lanatus has been recorded on abandoned plots of land in city suburbs (3). Velvet grass can tolerate a wide range of soil types and has been deliberately introduced to many areas for grazing or soil stabilization, and also accidentally introduced as a common contaminator of grass crop seed (2).
H. lanatus is considered invasive as it persists after introduction as a dominant habitat generalist that can monopolize water and nutrients, shade out seedlings of native plants, change litter composition, and alter soil chemistry, thereby significantly changing the plant composition of the invaded ecosystem (2). Velvet grass is also poisonous to mammals and listed as a noxious weed in North America (2).
Worldwide distribution 
The velvet grass H. lanatus is native throughout Europe and can be found worldwide in temperate climates of North America, South America, Africa, Australia, New Zealand, India, and Hawaii (1, 2). Peak airborne levels of velvet grass pollen have been recorded at the beginning of June in Vienna (Austria) and Berlin (Germany) (4), and from October to December in the Western Cape (South Africa) (5)
Route of Exposure
Inhalation of pollen from velvet grass can induce respiratory and cutaneous symptoms in sensitized individuals (4, 6-8)
Clinical Relevance
Pollen from velvet grass can induce respiratory and cutaneous symptoms in sensitized individuals such as allergic rhinitis, or hay fever (4, 7-9). A study in Berlin (Germany) demonstrated a highly significant relationship between daily concentrations of pollen species including H. lanatus and rhinitis symptoms of grass pollen allergy sufferers (p<0.016) (4). In Costa Rica, approximately 3 in 4 (77.7%) of 184 patients with allergic rhinitis showed positive skin prick tests to H. lanatus pollen (7). In Sweden, 89% of 133 individuals with seasonal allergic rhinitis had a positive intracutaneous reaction to H. lanatus grass pollen (referred to by the authors as ‘Yorkshire fog’) (8).
Molecular Aspects
Allergenic molecules
The following allergens and their molecular epitopes have been characterized from H. lanatus (10).
Name
Source
Biological Function
Molecular Weight
Grasses, Holcus lanatus, Plants, Poaceae, Velvet Grass (10)
Hol l 1, Hol l 1.0101, and Hol l 1.0102
A Group 1 protein, a beta-expansin (10, 11)
34 kDa(11, 12)
Hol l 2
A Group 2 protein with unknown function (10)
-
Hol l 4
A Group 4 protein, a Berberine bridge enzyme (10)
55-60 kDa (13)
Hol l 5, Hol l 5.0101, and Hol l 5.0201
A Group 5 enzyme, a ribonuclease (10)
30 kDa (11, 12)
Name
Source
Biological Function
Molecular Weight
In addition, H. lanatus pollen contains a proteinase inhibitor to bovine trypsin, comprising less than 1% of the total extractable pollen allergens, and recognized as an allergen by specific IgE antibodies in the serum of sensitized patients with pollinosis (14).
Cross-reactivity
Extensive IgE cross-reactivity has been established between velvet grass and fellow members of the subfamily Pooideae. Allergens from Group 1 and Group 5 appear to dominate the immune response to grass pollen extract, based on the prevalence of IgE antibody recognition among grass pollen-sensitized subjects (11).
Various authors have reported consistently high homology between Hol l 1 and Group 1 allergens from other Pooideae grasses. In a comparison of nucleotide sequences, there was approximately 90% sequence identity and similarity of Hol l 1 with Phl p 1 (Timothy grass, Phleum pratense), Lol p 1 (perennial ryegrass, Lolium perenne), and Pha a 1 (canary grass, Phalaris aquatica) (15); 95% homology and 93% identity with Phl p 1 in an analysis of the surface topographies of the allergens (16); and 88.1% sequence identity with Pha a 1 (17). Approximately 70% similarity has been demonstrated between Hol l 1 and Zea m 1 (maize, Zea mays) of the Poaceae subfamily Panicoideae (15), however no antigenic cross-reactivity was seen between Hol l 1 and Cyn d 1 (Bermuda grass, Cynodon dactylon) of the subfamily Chloridoideae (9).
The two isoforms rHol l 5.01 and rHol l 5.01 from H. lanatus are highly similar (range 64.6% to 83.0% sequence identity) to Group 5 allergens and their isoforms from other Pooideae homologs such as Phl p 5, Lol p 5, Pha a 5, and Poa p 5 (Kentucky bluegrass, Poa pratensis) (6). Hol l 5 harbors multiple discontinuous as well as continuous IgE-binding epitopes spread over the whole molecule, which were individually recognized by IgE antibodies from sera of 65 grass pollen allergic patients (18). Additionally, individual T cells directed against Timothy grass allergen Phl p 5.0101 cross-recognized corresponding epitopes of velvet grass, canary grass and Kentucky bluegrass (19).
More than 75% of grass pollen allergic patients produce specific IgE antibodies against Group 4 allergens (13). Inhibition studies of IgE antibody binding to Dac g 4 from orchard grass pollen (Dactylis glomerata) confirmed the presence of cross-reactive Group 4 allergens of 60 kDa in pollen from velvet grass, perennial ryegrass, Kentucky bluegrass, Timothy grass, rye (Secale cereale), tall fescue (Festuca elatior), field brome (Bromus arvensis), and barley (Hordeum sativum) (20). In another study, monoclonal antibodies to Phl p 4 from Timothy grass also bound strongly to Group 4 homologs in velvet grass, orchard grass, perennial ryegrass, and Kentucky bluegrass (13).
In a study of four patients with chronic cough, positive allergenicity to a 15 kDa excretory-secretory protein from Acanthamoeba was closely related to allergenicity to several species including velvet grass, willow tree, poplar, elm, oak, and cockroach (21). A study in the Cape Peninsula (South Africa) recorded positive radioallergosorbent test (RAST) to Timothy grass in 90% of grass-sensitive patients, even though Timothy grass does not occur in that location. This study suggested the responses probably represented cross-reactivity with other Pooideae grasses which do grow in that area, such as H. lanatus, L. perenne, annual meadow grass (Poa annua), sweet vernal grass (Anthoxanthum odoratum), cat grass (Avena sativa), and reed canary grass (Phalaris arundinacea) (5). An earlier study in Sweden reported that more than 85% of a cohort of 116 individuals with seasonal allergic rhinitis and positive skin reactions to H. lanatus (reported as ‘Yorkshire fog’ by the authors) were also positive to perennial ryegrass and Timothy grass (8).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:April 2022
References
  1. Gucker CL. Holcus lanatus 2008 [cited 2022 19.01.22]. Available from: https://www.fs.fed.us/database/feis/plants/graminoid/hollan/all.html.
  2. CABI. Holcus lanatus (common velvet grass) 2021 [cited 2022 19.01.22]. Available from: https://www.cabi.org/isc/datasheet/114824.
  3. Taketomi EA, Sopelete MC, de Sousa Moreira PF, de Assis Machado Vieira F. Pollen allergic disease: pollens and its major allergens. Braz J Otorhinolaryngol. 2006;72(4):562-7.
  4. Kmenta M, Bastl K, Berger U, Kramer MF, Heath MD, Pätsi S, et al. The grass pollen season 2015: a proof of concept multi-approach study in three different European cities. World Allergy Organ J. 2017;10(1):31.
  5. Potter PC, Berman D, Toerien A, Malherbe D, Weinberg EG. Clinical significance of aero-allergen identification in the western Cape. S Afr Med J. 1991;79(2):80-4.
  6. Schramm G, Bufe A, Petersen A, Schlaak M, Becker WM. Molecular and immunological characterization of group V allergen isoforms from velvet grass pollen (Holcus lanatus). Eur J Biochem. 1998;252(2):200-6.
  7. Riggioni O, Montiel M, Fonseca J, Jaramillo O, Carvajal E, Rosencwaig P, et al. [Type I hypersensitivity to graminea pollens by tribe, in allergic rhinitis patients]. Rev Biol Trop. 1994;42 Suppl 1:47-53, 18.
  8. Löfkvist T, Svensson G. Cutaneous reactions to pollen extracts in patients with hay fever. Special references to grass pollen recommended for treatment in fixed combination. Acta Allergol. 1975;30(2-3):96-105.
  9. Schumacher MJ, Griffith RD, O'Rourke MK. Recognition of pollen and other particulate aeroantigens by immunoblot microscopy. Journal of Allergy and Clinical Immunology. 1988;82(4):608-16.
  10. Allergome.org. Holcus lanatus 2021 [cited 2022 19.01.22]. Available from: http://www.allergome.org/script/search_step2.php.
  11. Andersson K, Lidholm J. Characteristics and immunobiology of grass pollen allergens. Int Arch Allergy Immunol. 2003;130(2):87-107.
  12. Schramm G, Petersen A, Bufe A, Schlaak M, Becker WM. Identification and characterization of the major allergens of velvet grass (Holcus lanatus), Hol l 1 and Hol l 5. Int Arch Allergy Immunol. 1996;110(4):354-63.
  13. Fahlbusch B, Müller WD, Rudeschko O, Jäger L, Cromwell O, Fiebig H. Detection and quantification of group 4 allergens in grass pollen extracts using monoclonal antibodies. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 1998;28(7):799-807.
  14. Berrens L, Marañón F. IgE-binding trypsin inhibitors in plant pollen extracts. Experientia. 1995;51(9-10):953-5.
  15. Schramm G, Bufe A, Petersen A, Haas H, Schlaak M, Becker WM. Mapping of IgE-binding epitopes on the recombinant major group I allergen of velvet grass pollen, rHol l 1. J Allergy Clin Immunol. 1997;99(6 Pt 1):781-7.
  16. Johansen N, Weber RW, Ipsen H, Barber D, Broge L, Hejl C. Extensive IgE cross-reactivity towards the Pooideae grasses substantiated for a large number of grass-pollen-sensitized subjects. Int Arch Allergy Immunol. 2009;150(4):325-34.
  17. Suphioglu C, Singh MB. Cloning, sequencing and expression in Escherichia coli of Pha a 1 and four isoforms of Pha a 5, the major allergens of canary grass pollen. Clin Exp Allergy. 1995;25(9):853-65.
  18. Schramm G, Bufe A, Petersen A, Haas H, Merget R, Schlaak M, et al. Discontinuous IgE-binding epitopes contain multiple continuous epitope regions: results of an epitope mapping on recombinant Hol l 5, a major allergen from velvet grass pollen. Clin Exp Allergy. 2001;31(2):331-41.
  19. Delong J, Till SJ, Roti M, Tan V, James EA, Robinson D, et al. Investigation of Group 5 Grass Allergen T Cell Cross-reactivity by Multi-tetramer Analysis. Journal of Allergy and Clinical Immunology. 2009;123(2):S125.
  20. Leduc-Brodard V, Inacio F, Jaquinod M, Forest E, David B, Peltre G. Characterization of Dac g 4, a major basic allergen from Dactylis glomerata pollen. J Allergy Clin Immunol. 1996;98(6 Pt 1):1065-72.
  21. Park HK, Park MK, Kim KU, Kang SA, Park SK, Ahn SC, et al. Evaluation of allergic sensitivity to Acanthamoeba allergen in patients with chronic cough. Allergy Asthma Proc. 2016;37(2):141-7.