Allergenic molecules
The following allergens have been characterized from A. sativa: (11)
· Ave s 1, a Group 1 grass allergen, an expansin
· Ave s 2, a Group 2 grass allergen with unknown biological function
· Ave s 36, a glutenin
· Ave s 4, a Group 4 grass allergen, a berberine bridge enzyme
· Ave s 5, a Group 5 grass allergen, a ribonuclease
· Ave s 7, a calcium-binding protein, polcalcin
· Ave s 11S, a legumin-like protein (11S Globulin)
· Ave s 12, a profilin
· Ave s 13, a Group 13 grass allergen, a polygalacturonase
Biomarkers of severity
Group 13 allergens specifically occur in pollen of the major grass subfamilies, and Group 13-specific IgE antibodies can be considered immunological markers for genuine grass pollen sensitization (12).
Cross-reactivity
More than 95% of patients allergic to grass pollens have IgE antibodies to Group 1 allergens, while 60% have IgE antibodies to Group 2/3 allergens, 70% to Group 4 allergens, 70-80% to Group 5 allergens, and 20% to profilins (13).
Extensive cross-reactivity among the different individual species of the genus Avena may be expected, and to a certain degree among members of the family Poaceae, especially in the subfamily Pooideae, as a result of the variable degree of cross-reactivity between allergens in Groups 1, 2, 4, and 5 (13-16).
A survey of 193 grass pollen-allergic patients from different countries noted that inhibition of IgE binding to pollen extracts prepared from species more closely related to timothy grass was higher (e.g. Anthoxanthum odoratum [sweet vernal grass]: 65%, A. sativa: 66%, Lolium perenne [rye grass]: 64%, Poa pratensis [meadow grass/Kentucky Bluegrass]: 66%, Secale cereale [cultivated rye]: 63%, and Triticum sativum [cultivated wheat]: 63%) than to extracts from more distantly-related species (Cynodon dactylon [Bermuda grass]: 47%, Phragmites australis [Australian reed]: 55%, and Zea mays [maize/corn]: 45%) (13). In this study, immunologically detectable Group 5 and Group 2 allergens were found in all species except for C. dactylon and Z. mays (13).
In a survey of proteins cross-reactive with Group 1, Group 4 and Group 9 allergens, numerous allergen cognate proteins were identified in pollen from ten agricultural species including barley (Hordeum vulgare), maize/corn (Z. mays), rye (S. cereale), triticale (xTriticosecale cereale), oat (A. sativa), canola (Brassica napus) and sunflower (Helianthus annus) (17).