Contact Us
Component
g215 Phl p 5b
g215 Phl p 5b Scientific Information
Type:
Component
Name; WHO/IUIS:
Phl p 5b
Route of Exposure:
Airway
Biological function:
Not confirmed, probably ribonuclease
Allergen code:
g215
Source Material:
Timothy grass (Phleum pratense) pollen
Other Names :
Phl p Vb, Group 5 grass allergen, Ag25
Summary
Phl p 5 is a group 5 pollen allergen of Timothy grass (Phleum pratnese).  Group 5 grass pollen allergens have been found in several members of the Pooidieae subfamily.  Phlp 5 is considered as one of the major “species-specific allergens” from timothy grass pollen. Approximately 65-90% of grass pollen sensitized individuals in temperate climate areas are found to be positive for Phl p 5.
Group 5 allergen of timothy grass has a molecular weight of 28-32 kilodalton (kDa) and consists of two isoforms: Group 5a and Group 5b with wide variation in IgE epitopes, which can explain its high allergenicity. Phl p 5b is smaller in size than 5a and has a molecular weight of 32 kDa. Variants of Phl p 5b of timothy grass reveal a 25-30% amino acid sequence divergence.
Phl p 5b is a major allergenic molecule of timothy grass pollen due to its structural characteristics that aid in effector cell activation. It is a key target of IgE antibodies among 80-95% of grass pollen allergic patients. Adults are found to be more sensitized to Phl p 5b compared to the pediatric population. Recombinant Phl p 5b or r Phlp 5b shows quite a similarity with natural Phl p 5b and also possesses homology to other Group 5 allergens.
Phl p 5b is responsible for causing allergic rhinitis (AR) and bronchial asthma in grass pollen allergic patients.  Phl p 5b has been shown to be cross-reactive with similar Group 5 allergens from several temperate grass species and also grain species 
Epidemiology
Worldwide distribution
Timothy grass has a widespread distribution in the temperate climate; however, it is the most common sensitizing pollen in Europe. Among the European population, the sensitization to the timothy grass pollen has been found to be as follows in some of the countries including Germany about 28.5%, United Kingdom (UK) about 26.9%, Sweden 24.2%, Amsterdam  19.5%, Belgium 18.5% with a median prevalence of 16.9%  (1). In Austrian students, a prevalence of 36% sensitization to timothy grass pollen was detected using allergen microarray analysis (2). Within the U.S., about 69% of the population is exposed to timothy grass (3). In Canada, the prevalence of sensitivity to timothy grass pollen usually varies between 20.2% and 23.3% with a median prevalence of 21.8% in patients with allergic rhino-conjunctivitis (ARC) (4).
Phl p 5b sensitization has been detected in the western region of the United States and  Canada (5). In a study with the Czech population, IgE sensitization to Phl p 5b was found to be less frequent in children than in adults with grass pollen allergy (59.8% vs. 79.1%; p= 0.023) (6, 7). One Italian study on the pediatric population detected a lower frequency of IgE sensitization to Phl p 5b compared to other timothy grass allergenic components: Phl p 1, Phl p 7, and Phl p 12 (7). Similar to these findings, a study in Portugal detected that among 41 patients with respiratory allergic symptoms of unknown etiology, 13 adults and 7 children had sensitization to Phl p 5b. Moreover, in this study, the adults showed increased median values for Phl p 5b than children (8). A German study determined 81% sensitization for Phl p 5b among adult patients allergic to timothy grass and suffering from ARC (9). Likewise in one Spanish study with 138 adult patients suffering from grass pollen associated rhinitis and /or asthma, 62.8% of patients showed sensitization towards Phl p5b (10).
Phl p 5a and Phl p 5b possess considerably different patterns of IgE binding. In fact, in a study with 150 sera, 100 of the sera showed IgE reactivity to rPhlp 5a and all these sera showed binding of IgE to rPhlp 5b. Moreover, exclusive reactivity with rPhl p 5b was found among 19 out of 150 sera indicating that Phl p 5b has at least one key epitope for IgE antibodies, which is probably not present in Phl p 5a (11). 
Environmental Characteristics
Source and tissue
The allergenic glycoprotein Phl p 5b is found to be positioned in the surface of the amyloplasts (starch granules) and the cytosol of the pollen grains (11).
Recombinant Phl p 5b (rPhl p 5b) is a non-glycosylated protein produced in an E. coli strain carrying a cloned cDNA encoding P. pratense allergen Phl p 5b or in P. pastoris (11).  rPhlp 5b shows a high similarity with natural Phl p 5b and also possesses homology to other Group 5 allergens (11, 12).
Clinical Relevance
Specific molecules
Phl p 5 (Group 5 allergen) of timothy grass has a molecular weight of 28-32 kilodalton (kDa) and consists of two isoforms: Group 5a and Group 5b (11, 13). Phl p 5b is smaller in size than 5a (14).
Phlp 5 is considered as one of the major “species-specific allergens” from timothy grass pollen (8). Phl p 5b does not contain any consensus site for N-linked glycosylation but has 17 O-linked glycosylation sites (11, 15). Variants of Phl p 5b of timothy grass reveal a 25-30% amino acid sequence divergence. The most common type of amino acid is alanine, whereas the amino acid cysteine, methionine, histidine, and tryptophan are rarely found in Phl p 5b (11).
Cross-reactive molecules
Phlp 5 is a heterogeneous group of proteins with isoforms a and b, having differences in primary sequences. Antibodies raised against Phl p 5 allergens failed to determine cross-reactive antigens outside of the subfamily Pooideae. In fact, Phl p 5b has been found to show cross-reactivity with some grasses (16).  
Molecular Aspects
Biochemistry
The molecular weight of Phl p 5b is around 32 KDa (14) and cDNA analysis reveals the presence of 260-300 amino acid residues in the protein. The isoelectric point (pI) of Group 5 allergens of timothy grass ranges between 4.2 to 9 and mostly neutral to acidic. The calculated pI of Phl p 5b is 5.99 (11). Studies revealed that Phl p 5b possesses ribonuclease activity in its C-terminal domain and may play important role in the defense mechanisms (11, 12).
X-ray crystallography and nuclear magnetic resonance spectroscopic analysis of N-terminal and C-terminal domains of Phl p 5b revealed that the protein forms a folded globular domain made up of an anti-parallel 4-helix bundle and the structure is quite similar to the structure of cytochrome c (11, 17). Phl p 5b is a dimeric protein; the dimers are joined covalently by a disulfide bridge present between two Cys205 residues. Phl p 5b usually dimerizes in solution (11).
Isoforms, epitopes, antibodies
According to some studies isoforms ‘a’ and ‘b’ of Phl p 5 are immunologically comparable, whereas according to some other studies, the two isoforms show variation in both the cellular and humoral immune responses (11).
Epitope mapping detected IgE epitopes belonging to the N-terminal domain of Phl p 5b. Moreover, epitopes have also been identified in the C-terminal domain of the allergen (18). Most of the Phl p 5-specific IgE epitopes are conformational (17). Binding of numerous IgE to the multiple independent epitopes of Phl p 5b and their tight association with the antigen attributes to its high allergenic potential (18).
Phl p 5b can accommodate several antibodies at largely independent sites and show high IgE reactivity (18). 
Cross-reactivity
The recombinant human IgE Fab had been found to cross-react strongly with Phl p 5  and other natural Group 5 allergen from different grass and corn species indicating that recombinant human IgE Fab is able to identify a cross-reactive epitope in Group 5 allergens from various grass and corn species (14, 18).  In fact, Phl p 5 shows cross-reactivity with other Group 5 allergens of the Pooideae subfamily of temperate grass and also corn species. Phl p 5 does not show cross-reactivity with the pollen of monocots (14).
Moreover, a recent study detected rabbit IgG antibodies raised against helminth Schistosoma mansoni soluble egg antigens (SmSEA) were cross-reactive with Phl p 5b and this was possibly due to the presence of similar glycans (CCDs) between helminths and plants (15).
Diagnostic Relevance
Disease Severity
Phl p 5b, the major Group 5 pollen allergen of timothy grass is responsible for causing AR and bronchial asthma in a large number of sensitized individuals globally (19). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: October  2020
References
  1. Newson RB, van Ree R, Forsberg B, Janson C, Lotvall J, Dahlen SE, et al. Geographical variation in the prevalence of sensitization to common aeroallergens in adults: the GA(2) LEN survey. Allergy. 2014;69(5):643-51.
  2. Pablos I, Wildner S, Asam C, Wallner M, Gadermaier G. Pollen Allergens for Molecular Diagnosis. Curr Allergy Asthma Rep. 2016;16(4):31.
  3. Lanknow R, Escalmel M, Jacobsen R, Hocker S, Coyne T, al. e. Grass Pollen Exposure in th Continental United States:Species Prevalence and Population Patterns. J Allergy Clin Immunol. 2012.
  4. Chan-Yeung M, Anthonisen NR, Becklake MR, Bowie D, Sonia Buist A, Dimich-Ward H, et al. Geographical variations in the prevalence of atopic sensitization in six study sites across Canada. Allergy. 2010;65(11):1404-13.
  5. Nolte M, Barber D, Maloney J, Li Z, Kaur A, Galan A, et al. Timothy specific IgE levels are associated with efficacy and safety of timothy grass sublingual immunotherapy tablet. Ann Allergy Asthma Immunol. 2015;115(6):509-15 e2.
  6. Sekerkova A, Polackova M, Striz I. Detection of Phl p 1, Phl p 5, Phl p 7 and Phl p 12 specific IgE antibodies in the sera of children and adult patients allergic to Phleum pollen. Allergol Int. 2012;61(2):339-46.
  7. Cipriani F, Mastrorilli C, Tripodi S, Ricci G, Perna S, Panetta V, et al. Diagnostic relevance of IgE sensitization profiles to eight recombinant Phleum pratense molecules. Allergy. 2018;73(3):673-82.
  8. Almeida E, Caeiro E, Todo-Bom A, Duarte A, Gazarini L. Sensitization to grass allergens: Phl p1, Phl p5 and Phl p7 Phl p12 in adult and children patients in Beja (Southern Portugal). Allergol Immunopathol (Madr). 2019;47(6):579-84.
  9. Darsow U, Brockow K, Pfab F, Jakob T, Petersson CJ, Borres MP, et al. Allergens. Heterogeneity of molecular sensitization profiles in grass pollen allergy--implications for immunotherapy? Clin Exp Allergy. 2014;44(5):778-86.
  10. Beitia JM, Lopez-Matas MA, Alonso A, Vega A, Mateo B, Cardenas R, et al. Allergenic profile to Phleum pratense and immunological changes induced after grass allergen-specific immunotherapy. Int Arch Allergy Immunol. 2014;165(1):9-17.
  11. Andersson K, Lidholm J. Characteristics and immunobiology of grass pollen allergens. Int Arch Allergy Immunol. 2003;130(2):87-107.
  12. Bufe A, Schramm G, Keown MB, Schlaak M, Becker WM. Major allergen Phl p Vb in timothy grass is a novel pollen RNase. FEBS Lett. 1995;363(1-2):6-12.
  13. Jung S, Estrella N, Pfaffl MW, Hartmann S, Handelshauser E, Menzel A. Grass pollen production and group V allergen content of agriculturally relevant species and cultivars. PLoS One. 2018;13(3):e0193958.
  14. Flicker S, Vrtala S, Steinberger P, Vangelista L, Bufe A, Petersen A, et al. A human monoclonal IgE antibody defines a highly allergenic fragment of the major timothy grass pollen allergen, Phl p 5: molecular, immunological, and structural characterization of the epitope-containing domain. J Immunol. 2000;165(7):3849-59.
  15. Igetei JE, El-Faham M, Liddell S, Schramm G, Doenhoff MJ. Antigenic cross-reactivity between Schistosoma mansoni and pollen allergens from the birch tree (Betula verrucosa) and Timothy grass (Phleum pratense): involvement of shared glycan epitopes and implications for the hygiene hypothesis. Int J Parasitol. 2018;48(5):345-57.
  16. Sharma A, Sharma N, Bhalla P, Singh M. Comparative and Evolutionary Analysis of Grass Pollen Allergens Using Brachypodium distachyon as a Model System. PLoS One. 2017;12(1):e0169686.
  17. Focke-Tejkl M, Campana R, Reininger R, Lupinek C, Blatt K, Valent P, et al. Dissection of the IgE and T-cell recognition of the major group 5 grass pollen allergen Phl p 5. J Allergy Clin Immunol. 2014;133(3):836-45 e11.
  18. Levin M, Rotthus S, Wendel S, Najafi N, Kallstrom E, Focke-Tejkl M, et al. Multiple independent IgE epitopes on the highly allergenic grass pollen allergen Phl p 5. Clin Exp Allergy. 2014;44(11):1409-19.
  19. Rajashankar K, Bufe A, Weber W, Eschenburg S, Lindner B, Betzel C. Structure of the functional domain of the major grass-pollen allergen Phlp 5b. Acta Crystallogr D Biol Crystallogr. 2002;58(Pt 7):1175-81.