Whole Allergen
i70 Fire ant
i70 Fire ant Scientific Information
Type:
Whole Allergen
Display Name:
Fire ant
Route of Exposure:
Injection of venom
Family:
Formicidae (3)
Species:
Solenopsis invicta
Latin Name:
Solenopsis invicta
Other Names:
Red Fire Ant, Red Imported Fire Ant (1)
Summary
Solenopsis invicta, the red imported fire ant, is one of the most notorious invasive ant species worldwide with a rapidly expanding geographical range, and occurs in high densities in urban, agricultural, and non-agricultural areas. Aggressive foragers, worker fire ants will rapidly swarm and repeatedly sting any human or animal with which they come into contact. In humans, injection of the venom results in the formation of a white fluid-filled pustule or blister at the site of the sting approximately one day after the attack, a pathognomonic symptom characteristic only of fire ants. Protein allergens within the venom can elicit reactions ranging from an immediate localized burning sensation with dermal wheal and flare responses, to painful large local reactions characterized by pruritus, erythema, and edema lasting 24‒72 hours. Serious systemic allergic reactions resulting in life-threatening anaphylaxis have been reported for between 0.6% to 16% of individuals stung by fire ants, resulting in at least 80 fatalities, most of which were caused by fewer than five stings. Four allergenic molecules have been characterized to date from S. invicta venom, with variable degrees of cross-reactivity to homologs in other Solenopsis species as well as with venom homologs from other Hymenoptera, insect, and arachnid species.
Allergen
Nature
Solenopsis invicta, the red imported fire ant, is one of the most notorious invasive ant species, listed as a major aggressive ant species worldwide and characterized as a “super pest” due to its rapid expansion rate and severe effects on economy, health, ecology, and lifestyle (4-6).
The fire ant gets its name from the characteristic intense burning pain felt by its sting, delivered by worker ants to subdue prey and repel larger competitors from food or other resources (4, 5, 7). Aggressive foragers, fire ants do not need to be provoked to attack and will rapidly swarm and sting any human or animal with which they come into contact (4-7). Bites can occur year-round but are most commonly reported in the summer months (7, 8), possibly due to higher concentrations of antigens in fire ant venom at this time (9, 10).
S. invicta should be distinguished from other local fire ant species such as the black imported fire ant (S. richteri) (5). S. invicta are dark reddish-brown with a black abdomen and range in length from 1.5 to 5 mm (polymorphic workers) to 9 mm (queens) (4).
Taxonomy
Taxonomic tree of Solenopsis invicta (Red Imported Fire Ant) (4)
Domain
Eukaryota
Kingdom
Metazoa
Phylum
Arthropoda
Subphylum
Uniramia
Class
Insecta
Order
Hymenoptera
Family
Formicidae
Genus
Solenopsis
Taxonomic tree of Solenopsis invicta (Red Imported Fire Ant) (4)
Environmental Characteristics
Living environment
S. invicta are omnivorous social insects occurring in high densities in urban, agricultural, and non-agricultural areas (4, 6, 9). Worker ants build a nest, care for the queen and brood (eggs, larvae, and pupae) ants, defend the colony, and forage outside the nest at soil surface temperatures between 12‒51°C (4). Foraging activity is temperature-dependent, with a drop in activity during cooler months and in shadier habitats (4).
Smaller nests may not be visible if they are constructed in soil or under objects such as timber or rocks, whereas dome-shaped mounds of soil up to 50 cm in diameter can be constructed in sunny or open areas with no cover such as lawns, gardens, parks, roadsides, schoolyards, and golf courses (4, 5, 9). A fully developed colony can contain between 200,000 to 400,000 individual ants (4). S. invicta breeds and spreads rapidly and can relocate a colony quickly if a nest is disturbed (4).
Of concern in terms of contact with humans, entire colonies of S. invicta can move into buildings or vehicles to seek favorable nesting sites, particularly during flooding or very hot dry conditions (4-6, 9). Additionally, S. invicta is attracted to electrical fields (11) and to pheromones released when other worker ants are electrocuted, and may construct nests in cable boxes, transformers, and other warm sheltered electrical equipment, which can cause mechanical failure and extensive damage to residential and commercial property (4, 6, 9).
Worldwide distribution 
S. invicta is native to central South America and, following accidental introduction, has become widely established across southern North America, the Caribbean, Asian mainland and island nations, Australia, and New Zealand (4, 5, 7). With a high potential for rapid colonization in a wide range of habitats (6, 12), areas considered at risk of invasion by S. invicta include northern South America and Central America, much of the region immediately surrounding the Mediterranean Sea, areas near the Black and Caspian Seas, and vulnerable island ecosystems worldwide (4, 12).
Key factors limiting the geographical range of S. invicta include low temperature and low levels of precipitation, however, reports of successful interbreeding between S. invicta and other ant species suggest distribution could expand if new hybrid ants display cold and/or drought resistance (4, 8, 11).
Clinical Relevance
The lower body is the most frequent site of a stinging attack by S. invicta (7, 13). Infants, the elderly, and individuals with limited mobility may be at particular risk of being stung by numerous fire ants if they cannot move away from the threat (4, 9, 12, 14, 15).
One sting from S. invicta delivers between 0.04‒0.11 µl of venom and 10‒100 ng of protein (9, 11), however, in practice multiple stings are often received as worker ants swarm together in an attack (12). The venom of S. invicta is composed of 95% water-insoluble cytotoxic alkaloids and 5% aqueous proteins (7, 11). In humans, injection of the alkaloid component results in the formation of a white fluid-filled pustule or blister at the site of the sting approximately one day after the attack, a pathognomonic symptom characteristic only of fire ants (4, 11). Protein allergens can elicit reactions ranging from an immediate localized burning sensation with dermal wheal and flare responses, to painful large local reactions characterized by pruritus, erythema, and edema lasting 24‒72 hours (6, 7, 9).
Anecdotal reports suggest some individuals may be stung hundreds of times without developing systemic symptoms (11, 16), however, for hypersensitized individuals, a single sting can elicit anaphylaxis with flushing, generalized urticaria, angioedema, laryngeal edema, bronchospasm, or loss of consciousness (6, 7). Serious systemic allergic reactions resulting in life-threatening anaphylaxis have been reported for between 0.6% to 16% of individuals stung by fire ants, resulting in at least 80 fatalities (9, 11, 13-15). A survey of 29,300 physicians in the United States reported 83 fatal and two near-fatal fire ant sting reactions, with most deaths caused by fewer than five stings (13).
Stings can also trigger serum sickness, nephrotic syndrome, seizures, and worsening of pre-existing cardiopulmonary disease (7, 9, 11, 17, 18). Allergic reactions to fire ant stings have been reported by previously sensitized individuals as well as by non-fire ant-sensitized subjects who have documented sensitivity to yellow jacket (Vespula spp.) venom (11).
In the south-eastern USA, over 40 million people live in areas infested by S. invicta and an estimated 14 million people (35%) are stung every year (4). In 1998 in the state of South Carolina alone, one survey of 1,286 medical practitioners estimated that 600,000 cases of imported fire ant attack resulted in 33,000 people seeking medical treatment (94 people per 19,000 population), of whom 660 (1.9 per 10,000 population) were treated for anaphylaxis (19). In Taiwan, a recent observational study of 3,819 people stung by S. invicta reported that 21.8% (n=834) developed wheal and flare reactions, 7.5% (n=288) sought medical care, 2.78% (n=106) exhibited anaphylactic shock, and 0.55% (n=21) developed severe cellulitis and urticaria (6).
Molecular Aspects
Allergenic molecules
The following allergens and molecular epitopes have been characterized to date from S. invicta venom (1):
Allergen
Type
Mass (kDa)
Sol i 1
Phospholipase A
38
Sol i 2
Group 2 ant venom with unknown function
15
Sol i 3
Antigen 5 (Ag5); SCP domain-containing protein
26
Sol i 4
Group 4 ant venom with unknown function
15.5
Allergen
Type
Mass (kDa)
Cross-reactivity
Variable degrees of cross-reactivity have been reported for individuals with venom from Solenopsis species as well as with venoms from other Hymenoptera (20).
In a cohort of 450 patients seeking medical treatment for insect allergy in West Bengal (India), skin prick tests (SPT) identified multiple sensitivities to a variety of insects including S. invicta (84.59%, n=313), common black ant (Lasius niger: 87.62%, n=324), honey bee (Apis cerana indica: 67.02%, n=248), mosquito (Aedes aegypti: 66.67%, n=247), and common wasp (Vespula vulgaris: 30.27%, n=112) [LAH20]. In this study, only 13% of patients showed single sensitivity towards fire ant only, while 45% of patients were cross-sensitized to black ant, fire ant, and honey bee (21).
In a survey of 49 patients who developed systemic allergic reactions to the imported fire ant, five individuals had associated hypersensitivity to other insects including deer fly (n=1), saddleback caterpillar (n=1), spider (n=1), and bee and wasp (n=2) (16). In another survey, 36% (4/11) patients with scorpion venom allergy (Centruroides vittatus) had a history of anaphylaxis with positive SPT to imported fire ant venom, and a further two individuals had a history of large local reactions (22).
Sol i 1 shares some immunologic cross-reactivity with phospholipase vespid venoms (20), whereas the structure of Sol i 2 is not related to any other known proteins (11) and does not cross-react with bee or vespid phospholipases (20). Sol i 3 shares between 44 to 50% identity with Ag5 molecules from yellow jacket (Vespula spp.), Dolichovespula spp., and hornet (Vespa spp.), but does not exhibit consistent immunologic cross-reactivity with vespid Ag5 allergens (11, 23, 24). Sol i 4 shares 35% identity with Sol i 2 but no immunologic cross-reactivity (11).
Reactions to stings from any fire ant are likely to cause sensitization to other fire ant species (23). Sol i 1 and Sol i 3 from S. invicta are highly cross-reactive with Sol r 1 and Sol r 3, respectively, from S. richteri (the black imported fire ant) (11, 23). In addition, Sol i 2 has 78.2% and 72.3% amino acid sequence homology with Sol r 2 (S. richteri) and Sol gem 2 (S. geminata, a tropical fire ant native to Thailand), respectively (23). 
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:April 2022
References
  1. allergome.org. Sol i [Venom] 2021 [cited 2022 18.01.22]. Available from: https://www.allergome.org/script/dettaglio.php?id_molecule=1974.
  2. LOINC. Red Imported Fire Ant IgE Ab [Units/volume] in Serum 2021 [cited 2022 18.01.22]. Available from: https://loinc.org/6117-6/.
  3. ITIS. Solenopsis invicta 2021 [cited 2022 18.01.22]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=154239#null.
  4. CABI. Solenopsis invicta(red imported fire ant) Wallingford, UK2021 [cited 2022 18.01.22]. Available from: https://www.cabi.org/isc/datasheet/50569.
  5. Collins L, Scheffrahn RH. Red imported fire ant University of Florida2001 [cited 2022 18.01.22]. Available from: https://entnemdept.ufl.edu/creatures/urban/ants/red_imported_fire_ant.htm.
  6. Liu Y-S, Huang S-A, Lin IL, Lin C-C, Lai H-K, Yang C-H, et al. Establishment and Social Impacts of the Red Imported Fire Ant, Solenopsis invicta, (Hymenoptera: Formicidae) in Taiwan. International journal of environmental research and public health. 2021;18(10):5055.
  7. Kruse B, Anderson J, Simon LV. Fire Ant Bites.  StatPearls. Treasure Island (FL): StatPearls Publishing Copyright © 2022, StatPearls Publishing LLC.; 2022.
  8. Tracy JM, Demain JG, Quinn JM, Hoffman DR, Goetz DW, Freeman TM. The natural history of exposure to the imported fire ant (<em>Solenopsis invicta</em>). Journal of Allergy and Clinical Immunology. 1995;95(4):824-8.
  9. Kemp SF, deShazo RD, Moffitt JE, Williams DF, Buhner WA, 2nd. Expanding habitat of the imported fire ant (Solenopsis invicta): a public health concern. J Allergy Clin Immunol. 2000;105(4):683-91.
  10. Hannan CJ, Jr., Stafford CT, Rhoades RB, Wray BB, Baer H, Anderson MC. Seasonal variation in antigens of the imported fire ant Solenopsis invicta. J Allergy Clin Immunol. 1986;78(2):331-6.
  11. Hoffman DR. Fire ant venom allergy. Allergy. 1995;50(7):535-44.
  12. Bonnefoy X, Kampen H, Sweeney K, World Health Organization. Regional Office for E. Public health significance of urban pests. World Health Organization. Regional Office for Europe; 2008.
  13. Rhoades RB, Stafford CT, James FK, Jr. Survey of fatal anaphylactic reactions to imported fire ant stings. Report of the Fire Ant Subcommittee of the American Academy of Allergy and Immunology. J Allergy Clin Immunol. 1989;84(2):159-62.
  14. deShazo RD, Kemp SF, deShazo MD, Goddard J. Fire ant attacks on patients in nursing homes: an increasing problem. Am J Med. 2004;116(12):843-6.
  15. Rupp MR, deShazo RD. Indoor fire ant sting attacks: a risk for frail elders. Am J Med Sci. 2006;331(3):134-8.
  16. Rhoades RB, Schafer WL, Schmid WH, Wubbena PF, Dozier RM, Townes AW, et al. Hypersensitivity to the imported fire ant: A report of 49 cases. Journal of Allergy and Clinical Immunology. 1975;56(2):84-93.
  17. Candiotti KA, Lamas AM. Adverse neurologic reactions to the sting of the imported fire ant. Int Arch Allergy Immunol. 1993;102(4):417-20.
  18. Swanson GP, Leveque JA. Nephrotic syndrome associated with ant bite. Tex Med. 1990;86(3):39-41.
  19. Caldwell ST, Schuman SH, Simpson WM, Jr. Fire ants: a continuing community health threat in South Carolina. J S C Med Assoc. 1999;95(6):231-5.
  20. Hoffman DR, Dove DE, Moffitt JE, Stafford CT. Allergens in Hymenoptera venom: XXI. Cross-reactivity and multiple reactivity between fire ant venom and bee and wasp venoms. Journal of Allergy and Clinical Immunology. 1988;82(5, Part 1):828-34.
  21. Laha A, Sarkar T, Dey D, Mondal P, Bhattacharya S, Moitra S, et al. Assessment of Hymenoptera and Non-Hymenoptera Insect Bite and Sting Allergy Among Patients of Tropical Region of West Bengal, India. Journal of Medical Entomology. 2019;57(1):1-7.
  22. Nugent JS, More DR, Hagan LL, Demain JG, Whisman BA, Freeman TM. Cross-reactivity between allergens in the venom of the common striped scorpion and the imported fire ant. J Allergy Clin Immunol. 2004;114(2):383-6.
  23. Potiwat R, Sitcharungsi R. Ant allergens and hypersensitivity reactions in response to ant stings. Asian Pac J Allergy Immunol. 2015;33(4):267-75.
  24. Hoffman DR. Allergens in Hymenoptera venom XXV: The amino acid sequences of antigen 5 molecules and the structural basis of antigenic cross-reactivity. Journal of Allergy and Clinical Immunology. 1993;92(5):707-16.