Whole Allergen
m227 Malassezia spp.
m227 Malassezia spp. Scientific Information
Type:
Whole Allergen
Display Name:
Malassezia spp.
Route of Exposure:
Contact
Family:
Malasseziaceae
Latin Name:
Malassezia spp.
Summary
Fungal species belonging to the genus Malassezia are part of the normal microbome of healthy human skin. More than a dozen species are identified, the species most commonly found on healthy human skin are M. sympodialis, M. globose and M. restricta. There is evidence of an association of hypersensitivity to Malassezia-specific allergen and atopic dermatitis. A similar association with allergic respiratory disease is also proposed. Over three dozen Malassezia specific allergens are recognized belonging to M. furfur, M. sympodialis and M. globose species. Most known cross-reactivity is seen with allergens of other fungal species. 
Allergen
Nature
Malassezia yeasts, formally known as Pityrosporum orbiculare/ovale, are yeast-like unicellular fungi that form part of the normal skin flora in man and many animal species (1, 2). With the exception of Malassezia pachydermatitis, all Malassezia species are lipid dependent (3), which explains a predilection for seborrheic skin sites such as the head and neck (4). The species most commonly found on human skin are M. sympodialis, M. globosa and M. restricta (1, 4).
Malassezia yeasts form part of the healthy dermal microbiome and comprise 1–22 % of the phylogenetic composition of the skin microflora (4), which is dependent on the area of the body (5). Malassezia spp. are the main eukaryotic microbial flora normally found on the skin (4).
Malassezia spp. have been implicated in the pathogenesis of a diverse range of dermatological diseases, including pityriasis versicolor, folliculitis, seborrheic dermatitis, atopic dermatitis (AD) and psoriasis (6). An etiological role of Malassezia spp. in respiratory allergic disease, such as allergic rhinitis and asthma has also been proposed (7). However, other authors have suggested the pathophysiology of Malassezia in atopic disease is limited to allergic skin disease only (1, 2). 
Taxonomy
Taxonomic tree of Malassezia spp. (1)
Domain
Eukaryote
Kingdom
Fungi
Phylum
Basidiomycota
Subphylum
Ustilaginomycotina
Class
Malasseziomycetes
Subclass
Incertae sedis
Order
Malasseziales
Family
Malasseziaceae
Genus
Malassezia
Taxonomic tree of Malassezia spp. (1)
Epidemiology
Worldwide distribution 
There are over a dozen species of Malassezia, the majority of which are isolated from human skin. M. globosa is the dominant species worldwide (except for Canada and Indonesia), a degree of geographical variation of the prevalence of Malassezia species exists. In the early 1990s, prevalence of M. sympodialis in healthy human skin cultures was between 7–69%, with the highest prevalence found in Sweden and Canada. Whereas an Iranian study at the turn of the 20th century found M. furfur with a prevalence of 4–23% (3).
A high frequency of sensitization to Malassezia allergens in patients with head and neck type atopic dermatitis (55–68%) has been reported. However, it has also been suggested that the Malassezia population on lesional atopic dermatitis skin is reduced compared to healthy skin (1). 
Route of Exposure
Main 
The mechanism by which fungal infections of the skin can provoke hypersensitivity disease of the skin, or the respiratory tract is not clear (8). It is possible that the direct contact of allergens on the skin could trigger a Th2 cascade response and the link between atopic dermatitis and bronchial asthma, if proven, driven by cytokines derived from epithelial cells such as TSLP, Il-33 and IL-25 (7).
Clinical Relevance
A strong association of sensitization to Malassezia spp. allergens in patients with atopic dermatitis has been demonstrated in several studies (1). Casagrande and colleagues showed that circulating allergen specific IgE was associated with patients with atopic eczema. They reported that 52/97 patients with AE had detectable M. sympodialis-specific IgE, compared with 4/571 patients with other allergic diseases and 0/38 healthy controls (9).
Sweat may increase the growth rate of M. globosa (10), as the main allergen of M. globose, MGL_1304, has been identified in human sweat (11). This could explain how cholinergic urticaria, with a prevalence of up to 20% in young adult, is triggered by sweating, heat, and exercise (10).
Molecular Aspects
Allergenic molecules
Table based on data from Allergome.org (12)
Allergen name
Protein group (if known)
Size (kDa)
Malassezia species
Mal f 1
-
-
M. furfur
Mala f2
Peroxysomal membrane protein
21
M. furfur
Mala f3
Peroxysomal membrane protein
20
M. furfur
Mala f4
Mitochondrial malate dehydrogenase
35
M. furfur
Mala s 1
-
-
M. sympodialis
Mala s 5
(unknown) M. sympodialis
-
M. sympodialis
Mala s 6
Cyclophilin, M sympodialis
-
M. sympodialis
Mala s 7
-
-
M. sympodialis
Mala s 8
-
-
M. sympodialis
Mala s 9
-
-
M. sympodialis
Mala s 10
Heat shock protein 70
86
M. sympodialis
Mala s 11
Manganese superoxide dismutase
23
M. sympodialis
Mala s 12
Glucose-methanol-choline (GML) oxidoreductase
67
M. sympodialis
Mala s 13
Thioredoxin
13
M. sympodialis
Mala sl 6
-
-
M. slooffiae
Mala y 11
-
-
M. yamatoensis
Mala y 6
-
-
M. yamatoenses
Allergen name
Protein group (if known)
Size (kDa)
Malassezia species
Almost twenty allergens specific to Malassezia spp. are recognized by the WHO/IUIS Allergen Nomenclature Sub-committee. The best understood Malassezia spp. allergens are those described for M. sympodialis, (Mala s 5, 6, 10 and 14) which have sequence similarity to human proteins. Mala 1 and 7 have not similarity with any known proteins and their function remains undetermined. Mala s 11 has high sequence homology to the human mitochondrial enzyme manganese superoxide dismutase, as well as the homologous Aspergillus fumigatus enzyme (2).
A central European study found that a high proportion of patients with AD (24%) had elevated specific IgE for Mala s 1 (7). Furthermore, the same study described how sensitization of Mala s 11, along with Mala s 6, a cyclophilin, was associated with moderate-to-severe manifestations of disease. Mala s 6 sensitization was also associated with severe allergic rhinitis (7). Another study found strong correlation between positive skin prick test (SPT) and atopy patch test (APT) to recombinant Mala s 1, 5, 6 and 9 in patients with atopic eczema, with a reduced reaction observed for Mala s 7 (9).
A recent study using recombinant Mal f 1 on THP-1 derived dendritic cells has revealed a novel activation pathway, with significant elevation of TNF-α, Il-6 and Il-10 significantly elevated with no change in IL-12 levels (6). This indicates Mal f 1 triggers a TH22/TH17 pathway separate from the classic IgE-dependent Th2 pathway and could give an explanation why treatment targeting the Th2 pathway has reduced efficacy.
MGL_1304 is major allergen in human sweat and is secreted by M. globosa. One study showed MGL­_1304 triggered the highest histamine release activity from basophils of patients with allergic dermatitis and cholinergic urticaria who were challenged with Malassezia spp., antigens (11). In a further study, supMGL_1304 was strongly associated with eczema and total IgE, but not with sensitization to common airborne/inhalant allergens. The authors suggested that this supports hypothesis that a defective skin barrier enhances antigen uptake and sensitization (10).
Cross-reactivity
Malassezia specific allergens contain proteins with high sequence similarity to proteins from non-Malassezia genera (1). This is consistent with the general observation that the majority of fungal allergens have extensive cross-reactivity with fungi that are taxonomically unrelated (7). Consequently, Mala f 2 and 3 (peroxisomal membrane protein, or PMP), Mala s 6 (cyclophiline), Mala s 10 (Heat Shock Protein 70), Mala s 11 (MnSOD) and Mala s 13 (thioredoxin) all have the potential for cross-reactivity (1). The latter also has similarity with human proteins (7). 
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:February 2022
References
  1. Fukutomi Y, Taniguchi M. Sensitization to fungal allergens: Resolved and unresolved issues. Allergol Int. 2015;64(4):321-31.
  2. Gaitanis G, Magiatis P, Hantschke M, Bassukas ID, Velegraki A. The Malassezia genus in skin and systemic diseases. Clin Microbiol Rev. 2012;25(1):106-41.
  3. Nowicka D, Nawrot U. Contribution of Malassezia spp. to the development of atopic dermatitis. Mycoses. 2019;62(7):588-96.
  4. Glatz M, Bosshard PP, Hoetzenecker W, Schmid-Grendelmeier P. The Role of Malassezia spp. in Atopic Dermatitis. J Clin Med. 2015;4(6):1217-28.
  5. Oh J, Byrd AL, Deming C, Conlan S, Barnabas B, Blakesley R, et al. Biogeography and individuality shape function in the human skin metagenome. Nature. 2014;514(7520):59-64.
  6. Li L, Wang S, Zou Z, Tao A, Ai Y. Activation profile of THP-1 derived dendritic cells stimulated by allergen Mal f 1 beyond its IgE-binding ability. Int Immunopharmacol. 2018;62:139-46.
  7. Celakovska J, Vankova R, Bukac J, Cermakova E, Andrys C, Krejsek J. Atopic Dermatitis and Sensitisation to Molecular Components of Alternaria, Cladosporium, Penicillium, Aspergillus, and Malassezia-Results of Allergy Explorer ALEX 2. J Fungi (Basel). 2021;7(3).
  8. Goldman DL, Huffnagle GB. Potential contribution of fungal infection and colonization to the development of allergy. Med Mycol. 2009;47(5):445-56.
  9. Fischer Casagrande B, Flückiger S, Linder MT, Johansson C, Scheynius A, Crameri R, et al. Sensitization to the Yeast Malassezia Sympodialis Is Specific for Extrinsic and Intrinsic Atopic Eczema. Journal of Investigative Dermatology. 2006;126(11):2414-21.
  10. Altrichter S, Schumacher P, Alraboni O, Wang Y, Hiragun M, Hide M, et al. Sensitization against skin resident fungi is associated with atopy in cholinergic urticaria patients. Clinical and Translational Allergy. 2020;10(1):18.
  11. Hiragun T, Hiragun M, Ishii K, Kan T, Hide M. Sweat allergy: Extrinsic or intrinsic? J Dermatol Sci. 2017;87(1):3-9.
  12. allergome.org. Malassezia 2021 [cited 2021 16.12.21]. Available from: https://allergome.org/script/search_step2.php.