Whole Allergen
m9 Fusarium proliferatum
m9 Fusarium proliferatum Scientific Information
Type:
Whole Allergen
Display Name:
Fusarium proliferatum
Route of Exposure:
Inhalation
Family:
Nectriaceae
Species:
proliferatum
Latin Name:
Fusarium proliferatum/F.moniliforme
Other Names:
Fusarium moniliforme; Gibberella fujikuroi; soy bean root rot
Summary
Fusarium proliferatum (Gibberella fujikuroi) is a ubiquitous mold of the order Hypocreales, mostly found in plant materials and soil worldwide. It shows a variety of phenotypes and is a known plant pathogen and producer of mycotoxins. The particles causing allergic reactions are spores found in the environment.
F. proliferatum hypersensitivity may present with symptoms of respiratory allergy such as asthma or rhinitis; molds of the Fusarium genus are also able to cause opportunistic disease. Cross reactivity with other molds, especially if taxonomically close, may occur.
Allergen
Nature
The genus Fusarium is a group of ubiquitous fungi found in soil and water environments (1), very common in warm and tropical regions (2). These fungi are well known saprophytes but also plant pathogens (1), with the ability to produce a large number of mycotoxins. The mycotoxins produced by Fusarium spp. including F. proliferatum are (among others) deoxyvalenol (DON), zearalenone (ZON), T-2 toxin, fumonisins B1, B2 and B3, beauvericin and fusaproliferin (3-9). Mycotoxins contaminate crops and have serious effects on the feed industry as they decrease yield and quality of the crop (4, 5, 9); fumonisin in particular has the potential to cause disease in animals and humans (10-12).
Fusarium proliferatum is the anamorph (asexual form) of the fungus G. fujikuroi (teleomorph, the sexual form) (9). F. proliferatum is able to cause disease in a very diverse range of plant species including rice (3), soy bean (13), maize and wheat (14), barley (5), asparagus (7), ornamentals such as Sansevieria trifasciata (15), prairie grasses (6), garlic (9), fig, onion, palm and pine (8).
Fusarium spp. Colonies are fast growing, variable in color, with an aerial mycelium taking a felt-like appearance. Conidia (a type of spore) are produced; depending on the species, macro- and microconidia are produced; these vary in shape and size. F. proliferatum can produce microconidial chains (2).
There is considerable overlap between peak fungal activity season and other allergens such as grass and weed pollens, often masking the pathology caused by fungal spores in multi-sensitized people (16). Fusarium spp. spores were detected at higher levels in the autumn and contributed on average to 3.03% of the total mold spores caught during a study spanning a 2-year period (17).
Taxonomy
Taxonomic tree of Fusarium genus (18)
Domain
Eukaryota
Kingdom
Fungi
Phylum
Ascomycota
Subphylum
Pezizomycotina
Class
Sordariomycetes
Order
Hypocreales
Family
Nectriaceae
Genus
Fusarium
Taxonomic tree of Fusarium genus (18)
Tissue
Spores (17).
Epidemiology
Worldwide distribution 
Hypersensitivity caused by Fusarium spp. has been described in several countries around the world, including India (17), Sweden (19), Malaysia (20), Greece (21) and Taiwan (22, 23).
Environmental Characteristics
Worldwide distribution 
Fusarium proliferatum has been identified worldwide (18).
Route of Exposure
Main 
Inhalation (7).
Detection
Main methods
Personal volumetric petri plate sampler (17)
Measures 
The concentrations of Fusarium spp. spores ranged between 51–132 CFU/m3 (17).
Clinical Relevance
There appears to be a scarcity of studies focusing on the specific allergic conditions caused by F. proliferatum. However, several studies have shown associations between Fusarium spp. and patients with a history of allergic conditions such as asthma, eczema, conjunctivitis and rhinitis (20-24). 
Other diseases
F. proliferatum is able to cause opportunistic disease; a case report showed that a 78-year old patient presenting with fever as the only symptom fungemia in (25). In two other reports F. proliferatum caused fungus ball sinusitis (a non-invasive form of fungal rhinosinusitis) in three patients (26, 27). Fusarium spp. were isolated from 5,321 cases of fungal keratitis; of these, 155 were caused by F proliferatum (28). The related species Fusarium napiforme caused hypersensitivity pneumonitis in a 17-year old patient; the mold was isolated from the home environment (29).
F. proliferatum was identified as one of the two most likely Fusarium species that cause fusariosis in European countries (12).
Diagnostics Sensitization
The presence of allergen-specific antibodies is usually determined by skin prick tests (SPTs) and serology for IgE (16, 21). In a study of 571 patients with a history of atopy, 42.5% had a positive SPT result to mold allergens, including Fusarium spp. (21). In another study using SPTs in 85 patients with a history suspicious of inhalant allergies, 23.5% tested positive to Fusarium spp. Of the patients testing positive to Fusarium spp., 15.3% were found to have concomitant allergic conditions such as asthma, eczema or allergic conjunctivitis (20)
Prevention and Therapy
Allergen immunotherapy
Immunotherapy is currently not recommended for patients allergic to molds, due to complexities of the allergens and patient co-allergies (16).
Prevention strategies 
Avoidance is difficult to achieve (16), due to the wide range of environments in which Fusarium spp. can be found (18).
Molecular Aspects
Allergenic molecules
Two allergens from F. proliferatum have been described; a transaldolase (37.5kDa), which showed IgE binding of 47% in serum of respiratory atopic patients (22). Another allergen was described as a vacuolar serine protease (36.5kDa), which was recognized by IgE from serum of patients with asthma and/or rhinitis (23).
Table adapted from Allergome.org (30).
Allergen
Type
Mass (kDa)
Fus p 4
Transaldolase
37.5
Fus p 9
Serine protease
36.5
Allergen
Type
Mass (kDa)
Cross-reactivity
Cross-reactivity has been demonstrated to other molds. In a study carried out on 668 serum samples from patients which who had previously recorded at least one IgE positivity to fungal antigens, associations were observed between the patterns of IgE sensitization and fungal phylogenetic relationships. Using a panel of 17 fungal extracts including F. proliferatum, some samples were only positive to one fungal species, whereas many were multi-sensitized. The results suggest that the associations are likely due to antigen cross-reactivity between fungal species, not uncommon in more closely related species (19). In another study, IgE from patients with a history of respiratory allergies (asthma and/or rhinitis) showed cross-reactivity between a F. proliferatum serine protease and a Penicillium spp. antigen (23).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:April 2022
References
  1. Weber RW, Levetin E. Allergen of the month--Fusarium. Ann Allergy Asthma Immunol. 2014;112(5):A11.
  2. Thrane U. FUSARIUM. In: Robinson RK, editor. Encyclopedia of Food Microbiology. Oxford: Elsevier; 1999. p. 901-6.
  3. Amatulli MT, Spadaro D, Gullino ML, Garibaldi A. Molecular identification of Fusarium spp. associated with bakanae disease of rice in Italy and assessment of their pathogenicity. Plant Pathology. 2010;59(5):839-44.
  4. Binder EM, Tan LM, Chin LJ, Handl J, Richard J. Worldwide occurrence of mycotoxins in commodities, feeds and feed ingredients. Animal Feed Science and Technology. 2007;137(3):265-82.
  5. Glenn AE. Mycotoxigenic Fusarium species in animal feed. Animal Feed Science and Technology. 2007;137:213-40.
  6. Leslie JF, Zeller KA, Logrieco A, Mulè G, Moretti A, Ritieni A. Species diversity of and toxin production by Gibberella fujikuroi species complex strains isolated from native prairie grasses in Kansas. Appl Environ Microbiol. 2004;70(4):2254-62.
  7. Logrieco AF, Doko B, Moretti A, Frisullo S, Visconti A. Occurrence of fumonisin B1 and B2 in Fusarium proliferatum infected asparagus plants. Journal of Agricultural and Food Chemistry. 1998;46:5201-4.
  8. Desjardins A, Moretti A. Biological and Chemical Complexity of Fusarium proliferatum. The Role of Plant Pathology in Food Safety and Food Security. 2010;3.
  9. Stanković S, Levic J, Petrovic T, Logrieco A, Moretti A. Pathogenicity and mycotoxin production by Fusarium proliferatum isolated from onion and garlic in Serbia. European Journal of Plant Pathology. 2007;118:165-72.
  10. Marasas WF, Riley RT, Hendricks KA, Stevens VL, Sadler TW, Gelineau-van Waes J, et al. Fumonisins disrupt sphingolipid metabolism, folate transport, and neural tube development in embryo culture and in vivo: a potential risk factor for human neural tube defects among populations consuming fumonisin-contaminated maize. J Nutr. 2004;134(4):711-6.
  11. Ross PF, Nelson PE, Richard JL, Osweiler GD, Rice LG, Plattner RD, et al. Production of fumonisins by Fusarium moniliforme and Fusarium proliferatum isolates associated with equine leukoencephalomalacia and a pulmonary edema syndrome in swine. Appl Environ Microbiol. 1990;56(10):3225-6.
  12. Ramos JF, Nucci M. Fusarium and Fusariosis.  Reference Module in Biomedical Sciences: Elsevier; 2021.
  13. Chang KF, Hwang SF, Conner RL, Ahmed HU, Zhou Q, Turnbull GD, et al. First report of Fusarium proliferatum causing root rot in soybean (Glycine max L.) in Canada. Crop Protection. 2015;67:52-8.
  14. Desjardins AE, Manandhar G, Plattner RD, Maragos CM, Shrestha K, McCormick SP. Occurrence of Fusarium Species and Mycotoxins in Nepalese Maize and Wheat and the Effect of Traditional Processing Methods on Mycotoxin Levels. Journal of Agricultural and Food Chemistry. 2000;48(4):1377-83.
  15. Kee YJ, Zakaria L, Mohd MH. Morphology, phylogeny and pathogenicity of Fusarium species from Sansevieria trifasciata in Malaysia. Plant Pathology. 2020;69(3):442-54.
  16. Twaroch TE, Curin M, Valenta R, Swoboda I. Mold allergens in respiratory allergy: from structure to therapy. Allergy Asthma Immunol Res. 2015;7(3):205-20.
  17. Kochar S, Ahlawat M, Dahiya P, Chaudhary D. Assessment of allergenicity to fungal allergens of Rohtak city, Haryana, India. Allergy Rhinol (Providence). 2014;5(2):56-65.
  18. CABI. Fusarium proliferatum Wallingford, UK2021 [cited 2022 24.01.22]. Available from: https://www.cabi.org/isc/datasheet/24684.
  19. Soeria-Atmadja D, Onell A, Borgå A. IgE sensitization to fungi mirrors fungal phylogenetic systematics. J Allergy Clin Immunol. 2010;125(6):1379-86.e1.
  20. Wan Ishlah L, Gendeh BS. Skin prick test reactivity to common airborne pollens and molds in allergic rhinitis patients. Med J Malaysia. 2005;60(2):194-200.
  21. Gonianakis MI, Neonakis IK, Gonianakis IM, Baritaki MA, Bouros D, Potamias G, et al. Mold allergy in the Mediterranean Island of Crete, Greece: a 10-year volumetric, aerobiological study with dermal sensitization correlations. Allergy Asthma Proc. 2006;27(5):354-62.
  22. Chou H, Wu K-G, Yeh C-C, Tai H-Y, Tam MF, Chen Y-S, et al. The Transaldolase, a Novel Allergen of Fusarium proliferatum, Demonstrates IgE Cross-Reactivity with Its Human Analogue. PLOS ONE. 2014;9(7):e103488.
  23. Yeh CC, Tai HY, Chou H, Wu KG, Shen HD. Vacuolar Serine Protease Is a Major Allergen of Fusarium proliferatum and an IgE-Cross Reactive Pan-Fungal Allergen. Allergy, asthma & immunology research. 2016;8(5):438-44.
  24. Pumhirun P, Towiwat P, Mahakit P. Aeroallergen sensitivity of Thai patients with allergic rhinitis. Asian Pac J Allergy Immunol. 1997;15(4):183-5.
  25. Dananché C, Cassier P, Sautour M, Gautheron N, Wegrzyn J, Perraud M, et al. Fungaemia caused by Fusarium proliferatum in a patient without definite immunodeficiency. Mycopathologia. 2015;179(1-2):135-40.
  26. Hashemi SJ, Ardehali MM, Rezaie S, Zibafar E, Shoar MG, Rezaei-Matehkolaei A, et al. A Case of Fungus Ball-Type Pansinusitis Due to Fusarium proliferatum. Mycopathologia. 2015;180(3-4):251-5.
  27. Radulesco T, Varoquaux A, Ranque S, Dessi P, Michel J, Cassagne C. Maxillary fungus balls due to Fusarium proliferatum. J Mycol Med. 2019;29(1):59-61.
  28. Sun S, Lui Q, Han L, Ma Q, Siyu H, Li X, et al. Identification and Characterization of Fusarium proliferatum, a New Species of Fungi that Cause Fungal Keratitis. Scientific Reports. 2018;8.
  29. Lee SK, Kim SS, Nahm DH, Park HS, Oh YJ, Park KJ, et al. Hypersensitivity pneumonitis caused by Fusarium napiforme in a home environment. Allergy. 2000;55(12):1190-3.
  30. allergome.org. Fusarium proliferatum 2021 [cited 2022 24.01.22]. Available from: http://www.allergome.org/script/search_step2.php.